Skip to Content
Merck
All Photos(1)

Documents

SAB2502119

Sigma-Aldrich

Anti-VEGFA antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

MGC70609, MVCD1, VEGF, VPF, vascular endothelial growth factor A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

canine, pig, human

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VEGFA(7422)

Immunogen

Peptide with sequence C-RIKPHQGQHIGE, from the internal region of the protein sequence according to NP_001020537.2; NP_003367.4; NP_001020538.2; NP_001020539.2; etc.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena Eremeeva et al.
Nucleic acids research, 47(10), 4927-4939 (2019-04-11)
Biomedical applications of nucleic acid aptamers are limited by their rapid degradation in biological fluids and generally demand tedious post-selection modifications that might compromise binding. One possible solution to warrant biostability is to directly evolve chemically modified aptamers from xenobiotic
Fan Shen et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 114, 108787-108787 (2019-03-30)
CEMIP is a cell migration-inducing protein that is closely associated with carcinogenesis. However, the function of CEMIP has not been reported in ovarian cancer. Here we show that CEMIP expression level in ovarian cancer tissues was higher than that in
Wei Chen et al.
Biology of reproduction, 100(1), 268-280 (2018-08-08)
Controlled ovarian hyperstimulation (COH) impairs the synchronized development of endometrium and embryo, resulting in the failure of embryo implantation. Here, we investigated what effects electroacupuncture had on embryo implantation in COH rats. Female rats were randomly assigned to four groups:
Marie Wolf et al.
Scientific reports, 9(1), 11579-11579 (2019-08-11)
Following corneal injury, coordinated cellular and protein interactions occur at the wound site to restore tissue homeostasis. Regulation of this response is required to prevent the development of chronic inflammation, abnormal neovascularization, and fibrosis. The chemokine CCL2 and its primary
Baohui Xu et al.
Arteriosclerosis, thrombosis, and vascular biology, 39(8), 1652-1666 (2019-07-12)
We examined the pathogenic significance of VEGF (vascular endothelial growth factor)-A in experimental abdominal aortic aneurysms (AAAs) and the translational value of pharmacological VEGF-A or its receptor inhibition in aneurysm suppression. Approaches and Results: AAAs were created in male C57BL/6J

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service