Skip to Content
Merck
All Photos(1)

Key Documents

SAB2501155

Sigma-Aldrich

Anti-Tyrosine Hydroxylase Antibody

goat polyclonal

Synonym(s):

Anti-TYH, tyrosine hydroxylase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Anti-TH (C-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, canine, human

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TH(7054)

General description

Tyrosine hydroxylase (TH) is encoded by the gene mapped to human chromosome 11p15.5. The N-terminal end of the protein plays a vital role in maintaining intracellular stability of the enzyme. Tyrosine hydroxylase is expressed in various tissues such as brain, adrenal medulla, and sympathetically innervated tissues. The enzyme is characterized with a regulatory domain (R), a catalytic domain (C) at N-terminal end, and a coiled-coil domain at the C-terminal end.

Immunogen

Peptide with sequence CVQDELDTLAHAL, from the C Terminus of the protein sequence according to NP_954986.2; NP_000351.2; NP_954987.2.

Biochem/physiol Actions

Tyrosine hydroxylase (TH) specifically catalyzes the hydroxylation of the amino acid L-tyrosine to 3, 4-dihydroxy-L-phenylalanine (L-DOPA). TH is a rate-limiting enzyme involved in the synthesis of catecholamine, which is involved in several brain functions, such as attention, memory, cognition, and emotion. Phosphorylation of the N-terminal portion of tyrosine hydroxylase controls the degradation of this enzyme by the ubiquitin-proteasome pathway. Alteration in the expression of the gene results in the pathogenesis of Parkinson′s disease (PD) and point mutation in the gene coding for TH leads to Segawa′s syndrome.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A point mutation in the tyrosine hydroxylase gene associated with Segawa's syndrome.
Ludecke B
Human Gene Therapy (1995)
Human tyrosine hydroxylase and insulin genes are contiguous on chromosome 11.
O'Malley KL and Rotwein P
Nucleic Acids Research (1988)
Tyrosine hydroxylase and regulation of dopamine synthesis.
Archives of Biochemistry and Biophysics (2011)
TYROSINE HYDROXYLASE. THE INITIAL STEP IN NOREPINEPHRINE BIOSYNTHESIS.
The Journal of Biological Chemistry (1964)
The function of tyrosine hydroxylase in the normal and Parkinsonian brain.
CNS & Neurological Disorders Drug Targets (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service