Skip to Content
Merck
All Photos(1)

Documents

SAB2500809

Sigma-Aldrich

Anti-PODXL antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Gp200, Anti-MGC138240, Anti-PCLP, Anti-Podocalyxin-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse, canine

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PODXL(5420)

General description

PODXL (podocalyxin like) is an anti-adhesive transmembrane protein which belongs to the sialomucin protein family. It is associated with cancer development and aggressiveness. The protein is expressed in various cells including epithelial cells of kidney glomeruli, vascular endothelial cells, hematopoietic stem cells and platelets. The gene encoding PODXL is localized on human chromosome 7q32.3.

Immunogen

Peptide with sequence C-DNLTKDDLDEEEDTH from the C Terminus of the protein sequence according to NP_001018121.1 ; NP_005388.2.

Biochem/physiol Actions

PODXL (podocalyxin like) acts as an anti-adhesion molecule in kidney open filtration pathways. It interacts with the actin-binding protein ezrin in cancer. Using ezrin and the bridging protein NHERF (Na+/H+) exchanger regulatory factor, it connects to the cytoskeleton to regulate cytoskeletal-related functions in tumor progression. In human, PODXL stimulates the cell adherence capacity to immobilized ligands and vascular endothelial cells in presence of integrins.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Podocalyxin increases the aggressive phenotype of breast and prostate cancer cells in vitro through its interaction with ezrin.
Susana L
Cancer Research (2007)
Genome wide analysis of novel copy number variations duplications/deletions of different epileptic patients in Saudi Arabia
Muhammad M
BMC Genomics (2015)
Li Ling et al.
International journal of molecular medicine, 41(5), 2704-2714 (2018-02-14)
Diabetic nephropathy (DN) is one of the most common complications associated with type I and II diabetes mellitus. Long noncoding RNAs (lncRNAs) have been implicated in various physiological and pathological processes, and recent evidence has demonstrated that they are involved in the process
Cheng-Wei Lin et al.
International journal of oncology, 45(2), 710-718 (2014-05-14)
Metastasis-mediated death remains a major challenge in cancer treatment due to the lack of identifiable biomarkers for early diagnosis. Identifying tumor-specific biomarkers is critical for the development of diagnostic and therapeutic tools. In the present study, we found that podocalyxin-like

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service