Skip to Content
Merck
All Photos(2)

Key Documents

SAB2500042

Sigma-Aldrich

Anti-AIF1/IBA1 (Isoform 3) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-AIF-1, Anti-Allograft inflammatory factor 1, Anti-DASS-82G15.2, Anti-IBA1, Anti-IRT-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AIF1(199)

General description

AIF1 (allograft inflammatory factor 1) is a calcium-binding, cytoplasmic protein that has 143-amino acids. It is also known as IBA1 (ionized calcium-binding adapter molecule 1). It is expressed in the cells of the monocyte/macrophage lineage, as well as in microglial and dendritic cells. It is located on human chromosome 6p21.3.

Immunogen

Peptide with sequence C-NKQFLDDPKYSSDED from the internal region of the protein sequence according to NP_001614.3.

Application

Anti-AIF1/IBA1 (Isoform 3) antibody produced in goat has been used in western blotting and immunofluorescence.

Biochem/physiol Actions

AIF1 (allograft inflammatory factor 1) is an essential factor of macrophage chemotaxis. Expression of AIF1 regulates the proliferation of vascular smooth muscle cells by G-CSF (cytokine granulocyte-colony stimulating factor) autocrine expression.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tumor Evolution of Glioma-Intrinsic Gene Expression Subtypes Associates with Immunological Changes in the Microenvironment.
Wang Q, et al.
Cancer Cell, 32(1), 42-56 (2017)
Dorsal root ganglia in Friedreich ataxia: satellite cell proliferation and inflammation.
Arnulf H K, et al.
Acta Neuropathologica Communications, 4(1), 46-46 (2016)
Azhaar Ahmad Ashraf et al.
Frontiers in aging neuroscience, 16, 1393351-1393351 (2024-06-05)
Iron dyshomeostasis and neuroinflammation, characteristic features of the aged brain, and exacerbated in neurodegenerative disease, may induce oxidative stress-mediated neurodegeneration. In this study, the effects of potential priming with mild systemic iron injections on subsequent lipopolysaccharide (LPS)-induced inflammation in adult
dolescent binge alcohol exposure affects the brain function through mitochondrial impairment
Tapia-Rojas C, et al.
Molecular Neurobiology, 55, 4473-4491 (2018)
Inhibition of AIF-1 expression by constitutive siRNA expression reduces
macrophage migration, proliferation, and signal transduction initiated by
atherogenic stimuli.
Ying T, et al.
American Journal of Physiology. Cell Physiology (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service