Skip to Content
Merck
All Photos(3)

Key Documents

SAB2108757

Sigma-Aldrich

Anti-SLC10A1

affinity isolated antibody

Synonym(s):

Anti- NTCP1, Anti-NTCP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

38 kDa

species reactivity

mouse, human, guinea pig, dog, rat

concentration

0.5-1 mg/mL

technique(s)

immunoblotting: suitable

accession no.

NM_003049

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC10A1(6554)

General description

The gene SLC10A1 (solute carrier family 10 member 1) is mapped to human chromosome 14q24.2. It is mainly expressed in the liver cells.

Immunogen

Synthetic peptide directed towards the middle region of human SLC10A1

Biochem/physiol Actions

SLC10A1 (solute carrier family 10 member 1) is a bile acid transporter. It plays an important role in the circulation of bile acids. Mutation in this gene is associated with hypercholanemia. SLC10A1 also plays an important role in the entry of HBV (Hepatitis B virus) and silencing of this gene inhibits HBV infection. It works as a receptor for preS1 domain in HBV.

Sequence

Synthetic peptide located within the following region: VFSLAMKGDMNLSIVMTTCSTFCALGMMPLLLYIYSRGIYDGDLKDKVPY

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fatemeh Alaei Faradonbeh et al.
Frontiers in physiology, 13, 859294-859294 (2022-04-08)
Multidrug resistance-associated protein 2 (Mrp2) mediates biliary secretion of anionic endobiotics and xenobiotics. Genetic alteration of Mrp2 leads to conjugated hyperbilirubinemia and predisposes to the development of intrahepatic cholestasis of pregnancy (ICP), characterized by increased plasma bile acids (BAs) due
Clinical and molecular study of a pediatric patient with sodium taurocholate cotransporting polypeptide deficiency.
Deng M, et al.
Experimental and Therapeutic Medicine, 12, 3294-3300 (2016)
Hepatitis B virus efficiently infects non-adherent hepatoma cells via human sodium taurocholate cotransporting polypeptide.
Okuyama-Dobashi K, et al.
Scientific Reports, 5, 17047-17047 (2015)
A genetic variant of the NTCP gene is associated with HBV infection status in a Chinese population.
Yang J, et al.
BMC Cancer, 16, 211-211 (2016)
The nuclear receptor FXR, but not LXR, up-regulates bile acid transporter expression in non-alcoholic fatty liver disease.
Aguilar-Olivos NE, et al.
Annals of Hepatology, 14, 487-493 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service