Skip to Content
Merck
All Photos(3)

Documents

SAB2108496

Sigma-Aldrich

Anti-VDAC1

IgG fraction of antiserum

Synonym(s):

Anti- VDAC-1, Anti-PORIN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

31 kDa

species reactivity

sheep, human, guinea pig, dog, bovine, rabbit, horse, mouse, rat

concentration

0.5-1 mg/mL

technique(s)

immunoblotting: suitable
immunohistochemistry: suitable

accession no.

NM_003374

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VDAC1(7416)

Related Categories

General description

The previously assigned protein identifier Q5FVE7 has been merged into P21796. Full details can be found on the UniProt database.

Immunogen

Synthetic peptide directed towards the C terminal region of human VDAC1

Biochem/physiol Actions

The voltage-dependent anion-selective channel 1 (VDAC1) functions as a channel in membranous structures for the outer mitochondrial membrane, the cell membrane, endosomes, caveolae, the sarcoplasmatic reticulum, synaptosomes, and post-synaptic density fraction. A major function of VDAC1 in the plasma membrane is that of a NADH(-ferricyanide) reductase that may be involved in the maintenance of cellular redox homeostasis.

Sequence

Synthetic peptide located within the following region: SAKVNNSSLIGLGYTQTLKPGIKLTLSALLDGKNVNAGGHKLGLGLEFQA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service