Skip to Content
Merck
All Photos(4)

Key Documents

SAB1410449

Sigma-Aldrich

Anti-ALOX5 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

5-LO, 5-LOX, 5LPG, LOG5, MGC163204

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 78 kDa

species reactivity

mouse, human

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALOX5(240)

General description

The gene encoding arachidonate 5-lipoxygenase (ALOX5) is localized on human chromosome 10q11.21 and has 13 introns. It spans a region of 71.9 kbp. The ALOX5 protein consists of 673 amino acids and has a molecular weight of 77.9 kDa. It possesses two domains, a regulatory C2-like domain that binds to calcium and a carboxy-terminal catalytic domain. ALOX5 is expressed in B cells, dendritic cells, mast cells, macrophages and polymorphonuclear leukocytes.
This gene encodes a member of the lipoxygenase gene family and plays a dual role in the synthesis of leukotrienes from arachidonic acid. The encoded protein, which is expressed specifically in bone marrow-derived cells, catalyzes the conversion of arachidonic acid to 5(S)-hydroperoxy-6-trans-8,11,14-cis-eicosatetraenoic acid, and further to the allylic epoxide 5(S)-trans-7,9-trans-11,14-cis-eicosatetrenoic acid (leukotriene A4). Leukotrienes are important mediators of a number of inflammatory and allergic conditions. Mutations in the promoter region of this gene lead to a diminished response to antileukotriene drugs used in the treatment of asthma and may also be associated with atherosclerosis and several cancers. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined. (provided by RefSeq)

Immunogen

ALOX5 (NP_000689.1, 1 a.a. ~ 674 a.a) full-length human protein.

Sequence
MPSYTVTVATGSQWFAGTDDYIYLSLVGSAGCSEKHLLDKPFYNDFERGAVDSYDVTVDEELGEIQLVRIEKRKYWLNDDWYLKYITLKTPHGDYIEFPCYRWITGDVEVVLRDGRAKLARDDQIHILKQHRRKELETRQKQYRWMEWNPGFPLSIDAKCHKDLPRDIQFDSEKGVDFVLNYSKAMENLFINRFMHMFQSSWNDFADFEKIFVKISNTISERVMNHWQEDLMFGYQFLNGCNPVLIRRCTELPEKLPVTTEMVECSLERQLSLEQEVQQGNIFIVDFELLDGIDANKTDPCTLQFLAAPICLLYKNLANKIVPIAIQLNQIPGDENPIFLPSDAKYDWLLAKIWVRSSDFHVHQTITHLLRTHLVSEVFGIAMYRQLPAVHPIFKLLVAHVRFTIAINTKAREQLICECGLFDKANATGGGGHVQMVQRAMKDLTYASLCFPEAIKARGMESKEDIPYYFYRDDGLLVWEAIRTFTAEVVDIYYEGDQVVEEDPELQDFVNDVYVYGMRGRKSSGFPKSVKSREQLSEYLTVVIFTASAQHAAVNFGQYDWCSWIPNAPPTMRAPPPTAKGVVTIEQIVDTLPDRGRSCWHLGAVWALSQFQENELFLGMYPEEHFIEKPVKEAMARFRKNLEAIVSVIAERNKKKQLPYYYLSPDRIPNSVAI

Biochem/physiol Actions

The gene ALOX5 (arachidonate 5-lipoxygenase) encodes a lipoxygenase that catalyzes the conversion of arachidonic acid (AA) and other fatty acids to bioactive compounds such as 5(S)-hydroxyeicosatetraenoic acid (5-HETE), leukotriene B4 (LTB4), and other leukotrienes. These leukotrienes and lipoxins function as mediators of inflammatory response. ALOX5 has been implicated in tumorigenesis and may serve as a potential therapeutic target and biomarker.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caixia Li et al.
Molecular medicine reports, 19(1), 727-733 (2018-11-06)
Adenomyosis is a common benign disease of women of childbearing age. The typical clinical features are prolonged menstruation, menorrhagia and ingravescent dysmenorrhea. In the present study, the severity of dysmenorrhea was assessed using the visual analogue scale system as follows:
Identification and Characterization of a New Protein Isoform of Human 5-Lipoxygenase.
Hafner AK, et al.
PLoS ONE, 11(11), e0166591-e0166591 (2016)
Jie Wu et al.
European journal of pharmacology, 859, 172484-172484 (2019-06-24)
Cerebral ischemia is one of the main causes of human neurological dysfunction. Baicalin (BC) and Geniposide (GP) and their combination (BC/GP) have an ameliorative effect on cerebral ischemia. Here, we use network pharmacology to predict the targets of BC, GP
Seong Eun Jin et al.
Molecules (Basel, Switzerland), 24(2) (2019-01-16)
Atopic dermatitis (AD) is a prevalent chronic inflammatory skin disease. The use of immunomodulatory corticosteroids in AD treatment causes adverse side effects. Therefore, novel natural anti-inflammatory therapeutics are needed. The aim of the present study was to investigate the anti-allergic
Katir K Patel et al.
Frontiers in immunology, 9, 2357-2357 (2018-10-31)
Background: Hepoxilins are biologically active metabolites of arachidonic acid that are formed through the 12-lipoxygenase pathway. Hepoxilin A3 is now known to be an important regulator of mucosal inflammation in response to infection by bacterial pathogens and was recently identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service