Skip to Content
Merck
All Photos(1)

Documents

SAB1408587

Sigma-Aldrich

Anti-IFI16 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

IFNGIP1, MGC9466, PYHIN2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~82 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... IFI16(3428)

Immunogen

IFI16 (AAH17059.1, 1 a.a. ~ 729 a.a) full-length human protein.

Sequence
MGKKYKNIVLLKGLEVINDYHFRMVKSLLSNDLKLNLKMREEYDKIQIADLMEEKFRGDAGLGKLIKIFEDIPTLEDLAETLKKEKLKVKGPALSRKRKKEVDATSPAPSTSSTVKTEGAEATPGAQKRKKSTKEKAGPKGSKVSEEQTQPPSPAGAGMSTAMGRSPSPKTSLSAPPNTSSTENPKTVAKCQVTPRRNVLQKRPVIVKVLSTTKPFEYETPEMEKKIMFHATVATQTQFFHVKVLNTSLKEKFNGKKIIIISDYLEYDSLLEVNEESTVSEAGPNQTFEVPNKIINRAKETLKIDILHKQASGNIVYGVFMLHKKTVNQKTTIYEIQDDRGKMDVVGTGQCHNIPCEEGDKLQLFCFRLRKKNQMSKLISEMHSFIQIKKKTNPRNNDPKSMKLPQEQSQLPNPSEASTTFPESHLRTPQMPPTTPSSSFFTKKSEDTISKMNDFMRMQILKEGSHFPGPFMTSIGPAESHPHTPQMPPSTPSSSFLTTLKPRLKTEPEEVSIEDSAQSDLKEVMVLNATESFVYEPKEQKKMFHATVATENEVFRVKVFNIDLKEKFTPKKIIAIANYVCRNGFLEVYPFTLVADVNADRNMEIPKGLIRSASVTPKINQLCSQTKGSFVNGVFEVHKKNVRGEFTYYEIQDNTGKMEVVVHGRLTTINCEEGDKLKLTCFELAPKSGNTGELRSVIHSHIKVIKTRKNKKDILNPDSSMETSPDFFF

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional Roles of Pattern Recognition Receptors That Recognize Virus Nucleic Acids in Human Adipose-Derived Mesenchymal Stem Cells.
Yu L
BioMed Research International, 2016 (2016)
Low-risk identification in multiple myeloma using a new 14-gene model.
Chen T
European Journal of Haematology, 89, 28-28 (2012)
Ravera Raffaella et al.
Experimental cell research, 293(2), 331-345 (2004-01-20)
Immunohistochemical analysis has demonstrated that the human IFI16 gene, in addition to the hematopoietic tissues, is highly expressed in endothelial cells and squamous stratified epithelia. In this study, we have developed a reliable HSV-derived replication-defective vector (TO-IFI16) to efficiently transduce
Mutsuko Ouchi et al.
Frontiers in bioscience : a journal and virtual library, 13, 236-239 (2007-11-06)
IFI16 is a member of the HIN-200 family (hematopoietic interferon-inducible nuclear antigens with 200 amino acid repeat) that contains a DNA binding domain, a transcriptional regulatory domain, DAPIN/PAAD domain associated with interferon (IFN) response and a binding domain for BRCA1
Nobuko Fujiuchi et al.
The Journal of biological chemistry, 279(19), 20339-20344 (2004-03-03)
IFI16 is a member of the PYRIN superfamily that has been implicated in BRCA1-mediated apoptosis and inflammation signaling pathways. Here we report that most breast cancer cell lines examined expressed decreased mRNA and protein levels of IFI16, although IFI16 is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service