Skip to Content
Merck
All Photos(5)

Documents

SAB1402541

Sigma-Aldrich

Monoclonal Anti-ACOT7, (N-terminal) antibody produced in mouse

clone 1E7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

ACH1, ACT, BACH, CTE-II, LACH, LACH1, MGC1126, hBACH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1E7, monoclonal

form

buffered aqueous solution

mol wt

antigen ~63.29 kDa

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACOT7(11332)

General description

This gene encodes a member of the acyl coenzyme family. The encoded protein hydrolyzes the CoA thioester of palmitoyl-CoA and other long-chain fatty acids. Decreased expression of this gene may be associated with mesial temporal lobe epilepsy. Alternatively spliced transcript variants encoding distinct isoforms with different subcellular locations have been characterized. (provided by RefSeq)

Immunogen

ACOT7 (AAH17365, 1 a.a. ~ 338 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MSGPDVETPSAIQICRIMRPDDANVAGNVHGGTILKMIEEAGAIISTRHCNSQNGERCVAALARVERTDFLSPMCIGEVAHVSAEITYTSKHSVEVQVNVMSENILTGAKKLTNKATLWYVPLSLKNVDKVLEVPPVVYSRQEQEEEGRKRYEAQKLERMETKWRNGDIVQPVLNPEPNTVSYSQSSLIHLVGPSDCTLHGFVHGGVTMKLMDEVAGIVAARHCKTNIVTASVDAINFHDKIRKGCVITISGRMTFTSNKSMEIEVLVDADPVVDSSQKRYRAASAFFTYVSLSQEGRSLPVPQLVPETEDEKKRFEEGKGRYLQMKAKRQGHAEPQP

Biochem/physiol Actions

ACOT7 (acyl-CoA thioesterase 7) is involved in fatty acid oxidation. It is a connecting link between metabolism of acyl-CoAs and cholesterol in neurons. Overexpression of this gene in macrophage cell line modulates the generation of prostaglandins D2 and E2, thereby making it a drug target in inflammatory diseases. ACOT7 has high specificity for arachido-258 noyl-CoA (AA-CoA). It might also be associated with mesial temporal lobe epilepsy (MTLE).

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sterol Regulatory Element-Binding Protein-2 modulates human brain acyl-CoA hydrolase gene transcription.
Takagi M
Molecular and Cellular Biochemistry, 275, 199-206 (2005)
Structural basis for recruitment of tandem hotdog domains in acyl-CoA thioesterase 7 and its role in inflammation.
Forwood JK
Proceedings of the National Academy of Sciences of the USA, 104, 10382-10387 (2007)
Functional and structural properties of mammalian acyl-coenzyme A thioesterases.
Kirkby B
Progress in Lipid Research, 49, 366-377 (2010)
Aberrant cytosolic acyl-CoA thioester hydrolase in hippocampus of patients with mesial temporal lobe epilepsy.
Yang JW
Amino Acids, 27, 269-275 (2004)
Alternative exon usage selectively determines both tissue distribution and subcellular localization of the acyl-CoA thioesterase 7 gene products.
Hunt MC
Cellular and Molecular Life Sciences, 64, 1558-1570 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service