Skip to Content
Merck
All Photos(3)

Key Documents

SAB1400679

Sigma-Aldrich

Anti-RBM15 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-FLJ12479, Anti-FLJ21943, Anti-MGC119584, Anti-OTT, Anti-OTT1, Anti-SPEN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RBM15(64783)

General description

Members of the SPEN (Split-end) family of proteins, including RBM15, have repressor function in several signaling pathways and may bind to RNA through interaction with spliceosome components (Hiriart et al., 2005 [PubMed 16129689]).[supplied by OMIM

Immunogen

RBM15 (AAI03661.1, 1 a.a. ~ 544 a.a) full-length human protein.

Sequence
MSHRAKLAMSGKIIIRNPIKIGYGKATPTTRLWVGGLGPWVPLAALAREFDRFGTIRTIDYRKGDSWAYIQYESLDAAHAAWTHMRGFPLGGPDRRLRVDFADTEHRYQQQYLQPLPLTHYELVTDAFGHRAPDPLRGARDRTPPLLYRDRDRDLYPDSDWVPPPPPVRERSTRTAATSVPAYEPLDSLDRRRDGWSLDRDRGDRDLPSSRDQPRKRRLPEESGGRHLDRSPESDRPRKRHCAPSPDRSPELSSSRDRYNSDNDRSSRLLLERPSPIRDRRGSLEKSQGDKRDRKNSASAERDRKHRTTAPTEGKSPLKKEDRSDGSAPSTSTASSKLKSPSQKQDGGTAPVASASPKLCLAWQGMLLLKNSNFPSNMHLLQGDLQVASSLLVEGSTGGKVAQLKITQRLRLDQPKLDEVTRRIKVAGPNGYAILLAVPGSSDSRSSSSSAASDTATSTQRPLRNLVSYLKQKQAAGVISLPVGGNKDKENTGVLHAFPPCEFSQQFLDSPAKALAKSEEDYLVMIIVRAKLVEQRMKIWNSKL

Biochem/physiol Actions

RBM15 (RNA binding motif protein 15) is an RTE (RNA transport element) binding factor majorly involved in the mRNA export. It plays a vital role in the NXF (Nuclear export factor) pathway. It directly binds to NXF1 and provides receptor for the RNA export element RTE. This conjugated form helps to enhance RTE-mediated expression. RBM15 has the ability to identify specific RTE RNA in vitro, which finally enhances export and expression of RTE-containing reporter mRNAs in vivo.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrei S Zolotukhin et al.
Nucleic acids research, 37(21), 7151-7162 (2009-09-30)
The conserved mRNA export receptor NXF1 (Mex67 in yeast) assembles with messenger ribonucleoproteins (mRNP) in the nucleus and guides them through the nuclear pore complex into the cytoplasm. The DEAD family RNA helicase Dbp5 is essential for nuclear export of
Susan Lindtner et al.
The Journal of biological chemistry, 281(48), 36915-36928 (2006-09-27)
Retroviruses/retroelements provide tools enabling the identification and dissection of basic steps for post-transcriptional regulation of cellular mRNAs. The RNA transport element (RTE) identified in mouse retrotransposons is functionally equivalent to constitutive transport element of Type D retroviruses, yet does not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service