Skip to Content
Merck
All Photos(1)

Documents

SAB1306342

Sigma-Aldrich

ANTI-LAMIN B1(C-TERMINAL) antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

LMN2, LMNB, LMNB1, Lamin-B1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

66408 Da

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LMNB1(4001)

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Noha A Gouda et al.
Antioxidants (Basel, Switzerland), 11(10) (2022-10-28)
Dipeptidyl peptidase-4 (DPP-4) inhibitors are reported to exhibit promising effects on several pathological processes associated with Parkinson's disease (PD). To explore its repositioning potential as an antiparkinsonian agent, we evaluated the effects of omarigliptin (OMG), a DPP-4 inhibitor recently approved
Maria Paola Bertuccio et al.
International journal of molecular sciences, 25(14) (2024-07-27)
Exposure to 2.45 GHz electromagnetic radiation (EMR) emitted from commonly used devices has been reported to induce oxidative stress in several experimental models. Our study aims to evaluate the efficacy of sulforaphane, a well-known natural product, in preventing radiation-induced toxic
Zhenzhen Li et al.
Frontiers in pharmacology, 9, 1519-1519 (2019-01-24)
NFATc4, a member from the Nuclear Factor of Activated T cells (NFATs) transcription factor family, plays a pivotal role in the development of cardiac hypertrophy. NFATc4 is dephosphorylated by calcineurin and translocated from the cytoplasm to the nucleus to regulate
Phuong Linh Nguyen et al.
International journal of molecular sciences, 22(16) (2021-08-28)
The c-Jun N-terminal kinases (JNKs) are implicated in many neuropathological conditions, including neurodegenerative diseases. To explore potential JNK3 inhibitors from the U.S. Food and Drug Administration-approved drug library, we performed structure-based virtual screening and identified azelastine (Aze) as one of
Gerile Tu et al.
Molecular medicine reports, 13(3), 2320-2326 (2016-01-20)
Retinal pigmented epithelial cell (RPE) oxidative stress is known to have a vital role in the etiology of age‑related macular degeneration (AMD). The present study aimed to investigate whether allicin, a natural product with antioxidant activity, was able to protect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service