Skip to Content
Merck
All Photos(2)

Key Documents

SAB1300251

Sigma-Aldrich

Anti-CCL19 (N-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCL19(6363)

General description

CCL19 (C-C motif chemokine ligand 19) gene is mapped in human chromosome 9p13.3. CCL19 is constantly expressed in secondary lymphoid tissue.
The gene for CCL19 is one of several CC cytokine genes clustered on the p-arm of chromosome 9. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. CCL19 may play a role in normal lymphocyte recirculation and homing. It also plays an important role in trafficking of T cells in thymus, and in T cell and B cell migration to secondary lymphoid organs. It specifically binds to chemokine receptor CCR7.

Immunogen

CCL19 (NP_006265, 11-41)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human CCL19.

Biochem/physiol Actions

CCL19 (C-C motif chemokine ligand 19) is an homeostatic chemokine, involved in adaptive immune system. CCL19 recruits monocytes, macrophages, antigen presenting dendritic cells and naïve T-cells to secondary lymphoid organs by activation of CCR7 (C-C motif chemokine receptor 7). It controls cell migration in inflammation reaction and dysregulated angiogenesis in many types of diseases. CCL19 is upregulated in human atherosclerotic lesions. CCL19 is involved in the development of various inflammatory disorders such as asthma, atherosclerosis and rheumatoid arthritis. CCL19 might also be associated with the pathogenesis of rickettsiae infection.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher T Veldkamp et al.
Biochemistry, 54(27), 4163-4166 (2015-06-27)
CCL19 and CCL21 are chemokines involved in the trafficking of immune cells, particularly within the lymphatic system, through activation of CCR7. Concurrent expression of PSGL-1 and CCR7 in naive T-cells enhances recruitment of these cells to secondary lymphoid organs by
Hong-Ren Yu et al.
Journal of cellular physiology, 230(9), 2120-2127 (2015-02-07)
Resistin may be an important link between obesity and diabetes. Recent studies have suggested an association between resistin and atherogenic processes. In addition, CCL19 is highly expressed in human atherosclerotic lesions. The interplays among resistin, CCL19, and shear stress in
Elisabeth Astrup et al.
BMC infectious diseases, 14, 70-70 (2014-02-11)
Based on their essential role in concerting immunological and inflammatory responses we hypothesized that the homeostatic chemokines CCL19 and CCL21 may play a pathogenic role in rickettsiae infection. Serum levels of CCL19 and CCL21 in patients with R. africae and
Albert Zlotnik et al.
Genome biology, 7(12), 243-243 (2007-01-05)
The human chemokine superfamily currently includes at least 46 ligands, which bind to 18 functionally signaling G-protein-coupled receptors and two decoy or scavenger receptors. The chemokine ligands probably comprise one of the first completely known molecular superfamilies. The genomic organization
Yang Qin et al.
BMC musculoskeletal disorders, 15, 316-316 (2014-09-28)
It is well-documented that both chemokine (C-C motif) ligand 19 (CCL19) and 21 (CCL21) mediate cell migration and angiogenesis in many diseases. However, these ligands' precise pathological role in ankylosing spondylitis (AS) has not been elucidated. The objective of this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service