Skip to Content
Merck
All Photos(3)

Key Documents

SAB1101510

Sigma-Aldrich

Anti-SMOX (151-165) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-C20orf16, Anti-PAO, Anti-SMO, Anti-Spermine oxidase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~56 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMOX(54498)

General description

Spermine oxidase (SMOX) gene mapped to human chromosome 20p13, is a flavin adenine dinucleotide (FAD)-dependent oxidase. It encodes nucleus and cytoplasmic specific splice variants.

Immunogen

synthetic peptide corresponding to amino acids 151-165 of human SMOX

Application

Anti-SMOX (151-165) antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

Spermine oxidase (SMOX) catabolizes spermine to spermidine, 3-aminopropanal and hydrogen peroxide. This catalytic activity of SMOX leads to apoptosis and DNA damage in gastric epithelial cells. It is highly expressed in H. pylori  infected gastritis tissues and in ulcerative colitis (UC). Induced SMOX expression in breast cancer using spermine analogs is regarded as an effective anticancer treatment regime. It is regarded as a key marker correlating chronic inflammation with epithelial carcinogenesis. Elevation in the SMO levels is observed in inflammatory bowel disease and is implicated in the immunopathogenesis of Citrobacter rodentium infection mediated colitis.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Increased polyamines as protective disease modifiers in congenital muscular dystrophy
Kemaladewi DU, et al.
Human Molecular Genetics, 27(11), 1905-1912 (2018)
Nuclear localization of human spermine oxidase isoforms-possible implications in drug response and disease etiology
Murray-Stewart T, et al.
The FEBS journal, 275(11), 2795-2806 (2008)
Increased expression and cellular localization of spermine oxidase in ulcerative colitis and relationship to disease activity
Hong SKS, et al.
Inflammatory bowel diseases, 16(9), 1557-1566 (2010)
Distinct immunomodulatory effects of spermine oxidase in colitis induced by epithelial injury or infection
Gobert AP, et al.
Frontiers in Immunology, 9, 1242-1242 (2018)
Spermine oxidase (SMO) activity in breast tumor tissues and biochemical analysis of the anticancer spermine analogues BENSpm and CPENSpm
Cervelli M, et al.
BMC Cancer, 10(1), 555-555 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service