Skip to Content
Merck
All Photos(1)

Key Documents

S9008

Sigma-Aldrich

Anti-Staphylococcal Enterotoxin B antibody produced in rabbit

fractionated antiserum, lyophilized powder

Synonym(s):

Rabbit Anti-SEB

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

Staphylococcus aureus

packaging

vial of 2 mL lyophilized antiserum

technique(s)

indirect ELISA: 1:150,000-1:250,000

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Staphylococcal enterotoxin B is a toxic bacterial superantigen. The staphylococcal enterotoxins family have high sequence similarity.

Immunogen

Staphylococcal enterotoxin B from Staphylococcus aureus.

Application

Anti-Staphylococcal Enterotoxin B antibody produced in rabbit has been used:
  • to conjugate with hollow gold nanosphere and as a component of lateral flow immunoassay strip
  • in antibody sandwich immunoassay
  • for coupling with magnetic beads for detection assay

Biochem/physiol Actions

Staphylococcal enterotoxin B (SEB) is a superantigen that binds to class II MHC molecules on antigen presenting cells. It stimulates large populations of T cells resulting in acute toxic shock syndrome. It is resistant to denaturation and causes outbreaks of food poisoning that includes emetic responses, diarrhea and GI inflammation.
Staphylococcal enterotoxin B compromises the immune system by triggering inflammation and cytokine release.

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.2.

Reconstitution

Reconstitute with 2 ml deionized water.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of a SERS-based lateral flow immunoassay strip for the rapid and sensitive detection of staphylococcal enterotoxin B
Hwang J, et al.
Nanoscale, 8(22), 11418-11425 (2016)
Lily Zhang et al.
Toxins, 5(10), 1859-1871 (2013-10-25)
The functional activity of superantigens is based on capacity of these microbial proteins to bind to both the β-chain of the T cell receptor (TcR) and the major histocompatibility complex (MHC) class II dimer. We have previously shown that a
Electrogenerated indium tin oxide-coated glass surface with photosensitive interfaces: Surface analysis
Konry T, et al.
Biosensors And Bioelectronics, 22(9-10), 2230-2236 (2007)
Antje Heit et al.
The Journal of experimental medicine, 214(7), 2139-2152 (2017-06-24)
Germinal center T follicular helper cells (GCTfh) in lymphatic tissue are critical for B cell differentiation and protective antibody induction, but whether GCTfh establish clonal derivatives as circulating memory T cells is less understood. Here, we used markers expressed on
Staphylococcal enterotoxins
Balaban N and Rasooly A
International Journal of Food Microbiology, 61(1), 1-10 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service