Skip to Content
Merck
All Photos(2)

Key Documents

S7401

Sigma-Aldrich

Salicylic acid

suitable for plant cell culture

Synonym(s):

2-Hydroxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2H
CAS Number:
Molecular Weight:
138.12
Beilstein:
774890
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.71

vapor density

4.8 (vs air)

Quality Level

vapor pressure

1 mmHg ( 114 °C)

Assay

≥99% (HPLC)

form

powder

technique(s)

cell culture | plant: suitable

bp

211 °C (lit.)

mp

158-161 °C (lit.)

solubility

ethanol: 50 mg/mL

density

1.44 g/cm3 at 20 °C

SMILES string

OC(=O)c1ccccc1O

InChI

1S/C7H6O3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H,(H,9,10)

InChI key

YGSDEFSMJLZEOE-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Salicylic acid (SA) is a phenolic compound produced in plants.

Application

Salicylic acid has been used:
  • to study its effects on glandular trichomes in Artemisia annua
  • to study its effects on DNA methylation in Taraxacum alatum andTaraxacum hemicyclum
  • to study its effects on parasite epidemics in Lolium arundinaceum

Biochem/physiol Actions

Salicylic acid (SA) plays a role in endogenous signaling for plant disease resistance. SA is also involved in mediating thermogenesis and symptom development.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

314.6 °F - closed cup

Flash Point(C)

157 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D'Maris Amick Dempsey et al.
The arabidopsis book, 9, e0156-e0156 (2012-02-04)
Salicylic acid (SA) has been shown to regulate various aspects of growth and development; it also serves as a critical signal for activating disease resistance in Arabidopsis thaliana and other plant species. This review surveys the mechanisms involved in the
Anders Kjær et al.
AoB PLANTS, 2012, pls018-pls018 (2012-07-27)
Glandular trichomes (GT) of Artemisia annua produce valuable compounds for pharmaceutical and industrial uses, most notably the anti-malarial artemisinin. Our aim was to find out whether the density, number and size of GT can be manipulated to advantage by environmental
Veronica Preite et al.
Ecology and evolution, 8(5), 3047-3059 (2018-03-14)
DNA methylation is one of the mechanisms underlying epigenetic modifications. DNA methylations can be environmentally induced and such induced modifications can at times be transmitted to successive generations. However, it remains speculative how common such environmentally induced transgenerational DNA methylation
E Benfeldt et al.
Acta dermato-venereologica, 79(5), 338-342 (1999-09-24)
Our aim was to simultaneously investigate 2 techniques for in vivo sampling of peripheral compartment pharmacokinetics after systemic administration of acetylsalicylic acid. Ten volunteers were given 2 g acetylsalicylic acid orally. Blood samples and dialysates from 4 microdialysis probes inserted
S Zaugg et al.
Journal of chromatography. B, Biomedical sciences and applications, 752(1), 17-31 (2001-03-20)
Acetylsalicylic acid (Aspirin) is rapidly metabolized to salicylic acid (salicylate) and other compounds, including gentisic acid and salicyluric acid. Monitoring of salicylate and its metabolites is of toxicological, pharmacological and biomedical interest. Three capillary electrophoresis (CE) methods featuring alkaline aqueous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service