Skip to Content
Merck
All Photos(1)

Key Documents

S5171

Sigma-Aldrich

Human Fibronectin

formulated with recombinant human fibronectin fragment III-C expressed in E. coli and fibronectin from human plasma, liquid, 2 mg/mL, suitable for cell culture

Synonym(s):

sFN

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.75

Product Name

Superfibronectin from human plasma, solution, BioReagent, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in E. coli

product line

BioReagent

form

solution

mol wt

~235-270 kDa

packaging

pkg of 5 mg

technique(s)

cell culture | mammalian: suitable

surface coverage

1 μg/cm2

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... FN1(2335)

Application

Superfibronectin from human plasma dramatically enhances the adhesive properties of fibronectin and suppresses cell migration. Improved results have been shown CHO cells, IMR-90 fibroblasts, T47D breast carcinoma cells, and UCLA-P lung carcinoma cells. Additional applications include epithelial cells, mesenchymal cells, neuronal cells, fibroblasts, neural crest cells, and endothelial cells. It is recommended for use as a cell culture substratum at 1 μg/cm2.

Components

Superfibronectin is formulated with recombinant, human fibronectin fragment III-C expressed in E. coli and fibronectin from human plasma. The recombinant fibronectin includes a new region, type III, cloned from human placenta cDNA, that is newly reported to participate in matrix formation by inducing spontaneous disulfide crosslinking of fibronectin molecules into multimers. The product resembles the in vivo matrix form of fibronectin.

Caution

Superfibronectin should be stored at 2-8°C and diluted immediately prior to use. Do not freeze this product.

Preparation Note

As supplied, this product is a 2 mg protein/mL solution in 0.05 M Tris buffered saline. Before coating a 96 well plate, it is recommended to dilute t he product with sterile PBS to a concentration of 5 μg/mL and add 100 μl/well. These plates should then be incubated for 2 hours at 37°C and washed twice with 100 μl of PBS. The source material has been tested for HBsAg and HIV antibodies.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gentle cell handling with an ultra-slow instrument: creep-manipulation of cells
Thielecke H, et al.
Microscopica Acta, 11(11), 1230-1241 (2005)
N Okada et al.
The Journal of biological chemistry, 272(43), 26978-26984 (1997-10-27)
The pathogenic Gram-positive bacterium Streptococcus pyogenes (group A streptococcus) binds to fibronectin via protein F. In this study, we have investigated the binding properties of protein F to various multimeric tissue forms of fibronectin that appear on cell surfaces and
Regulated GPCR trafficking to the plasma membrane: general issues and the CCR5 chemokine receptor example
Giornale di Psichiatria e di Neuropatologia (2012)
A Morla et al.
Nature, 367(6459), 193-196 (1994-01-13)
Fibronectin is an extracellular matrix protein that is important in development, wound healing and tumorigenesis. In the blood it is dimeric, but in tissues forms disulphide crosslinked fibrils. Here we show that a fragment from the first type-III repeat of
Florian Weinberg et al.
International journal of molecular sciences, 21(23) (2020-12-03)
The epidermal growth factor receptor HER2 is overexpressed in 20% of breast cancer cases. HER2 is an orphan receptor that is activated ligand-independently by homodimerization. In addition, HER2 is able to heterodimerize with EGFR, HER3, and HER4. Heterodimerization has been

Articles

Methods for lentiviral transduction of Jurkat cells were compared. Spinoculation was compared with overnight incubation with polybrene (hexadimethrine bromide) and fibronection-coated plates.

Methods for lentiviral transduction of Jurkat cells were compared. Spinoculation was compared with overnight incubation with polybrene (hexadimethrine bromide) and fibronection-coated plates.

Methods for lentiviral transduction of Jurkat cells were compared. Spinoculation was compared with overnight incubation with polybrene (hexadimethrine bromide) and fibronection-coated plates.

Methods for lentiviral transduction of Jurkat cells were compared. Spinoculation was compared with overnight incubation with polybrene (hexadimethrine bromide) and fibronection-coated plates.

See All

Protocols

Dilute fibronectin for cell attachment, varying per cell type. Coating protocol, products, and FAQs provided.

Dilute fibronectin for cell attachment, varying per cell type. Coating protocol, products, and FAQs provided.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service