Skip to Content
Merck
All Photos(2)

Key Documents

S4521

Sigma-Aldrich

Saponin from Quillaja sp.

Sapogenin content 20-35 %

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.32

description

non-ionic

composition

Sapogenin content, 20-35%

technique(s)

HPLC: suitable
protein quantification: suitable

sulfated ash

≤10%

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@H]2[C@@H](O[C@@H]([C@H]([C@@H]2O)O[C@@H]8O[C@@H]([C@H]([C@@H]([C@H]8O)O)O)CO)C(=O)O)O[C@@H]3[C@@](C4[C@@](C5[C@]([C@@]6(C[C@H]([C@]7([C@H]([C@@H](C(CC7C6=CC5)(C)C)OC(=O)\C(=C/C)\C)OC(=O)C)CO)O)C)(CC4)C)(CC3)C

InChI

1S/C55H86O24/c1-10-23(2)46(71)79-43-44(72-24(3)60)55(22-59)26(17-50(43,4)5)25-11-12-30-51(6)15-14-32(52(7,21-58)29(51)13-16-53(30,8)54(25,9)18-31(55)61)75-49-41(77-48-38(67)36(65)34(63)28(20-57)74-48)39(68)40(42(78-49)45(69)70)76-47-37(66)35(64)33(62)27(1

InChI key

AXNVHPCVMSNXNP-ZELRDNAQSA-N

General description

Saponins are steroid or triterpenoid glycosides. They are biosurfactants that have several uses in the food and cosmetic industry.

Application

Saponin from Quillaja bark has been used:
  • to stain fertilized oocytes
  • in the analysis of intracellular cytokine production
  • to permeabilize dendritic cells

Biochem/physiol Actions

Potent hemolytic when injected i.v.; surfactant that enhances penetration of proteins and other macromolecules through cell membranes; it also has been used as an adjuvant for vaccines.
Saponins solubilize the low density lipoprotein (LDL) fraction of cholesterol in micelles, thus lowering their concentration. Saponin from Quillaja bark is used as a flavoring agent in food and beverages. This saponin has been shown to enhance immune-cell proliferation in vitro.

Preparation Note

Purified to remove low molecular weight contaminants 

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Louis Lignereux et al.
Preventive veterinary medicine, 157, 70-77 (2018-08-09)
Contagious caprine pleuropneumonia (CCPP), caused by Mycoplasma capricolum subsp. capripneumoniae, has long been considered a goat-specific disease. Since 2007 there has been growing evidence that this disease can affect wild ungulates either kept in captivity or in the wild. In
R Martin-Ibañez et al.
Human reproduction (Oxford, England), 23(12), 2744-2754 (2008-08-22)
Human embryonic stem cells (hESCs) have potential use in clinical therapy and regenerative medicine. One of the major challenges regarding the application of these cells is the development of an efficient cryopreservation protocol, since current methods, which include slow-freezing-rapid thawing
Adjuvant effects of saponins on animal immune responses.
Rajput ZI
Journal of Zhejiang University. Science. B, 8(3) , 153-161 (2007)
Isolation of myeloid and plasmacytoid dendritic cells from human bronchoalveolar lavage fluid.
Tsoumakidou M
Immunology and Cell Biology, 84(3), 267-273 (2006)
Dynamic chromatin modifications characterise the first cell cycle in mouse embryos.
Santos F
Developmental Biology, 280(1), 225-236 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service