Skip to Content
Merck
All Photos(1)

Key Documents

S2770

Sigma-Aldrich

Sodium hydroxide solution

1.0 N, BioReagent, suitable for cell culture

Synonym(s):

Caustic Soda, Lye, NaOH, Soda

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352316
PubChem Substance ID:
NACRES:
NA.71

grade

reagent

vapor pressure

3 mmHg ( 37 °C)

sterility

sterile-filtered

product line

BioReagent

form

solution

concentration

1.0 N

technique(s)

cell culture | mammalian: suitable

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium hydroxide is a strong alkali. Its reaction with acid yields salt and water. Sodium hydroxide solution has many industrial applications including dyes and drugs. It has the ability to disintegrate lignin and hence is used in paper manufacturing. Sodium hydroxide has high melting of about 318°C.

Application

Sodium hydroxide solution has been used:
  • to neutralize collagen type I solution phosphate-buffered saline for its use in 3D cell culture technique
  • to dissolve phosphate chains in phosphate glass for optical characterisation
  • as a component of transfer medium used for cell culture
  • as a general titration base for cell culture applications.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Matrix dimensions, stiffness, and structural properties modulate spontaneous chondrogenic commitment of mouse embryonic fibroblasts
Fernandez MT
Tissue Engineering: Part A, 20(7-8), 1145-1155 (2013)
Sodium, 21-21 (2002)
Optical characterisation of hydroxide catalysed bonds applied to phosphate glass
Optifab, 10448, 1044825-1044825 (2017)
Isolation and culture of human endometrial epithelial cells and stromal fibroblasts
Chen JC and Roan NR
Bio-protocol, 5(20) (2015)
Konstantinos A Polyzos et al.
Cardiovascular research, 106(2), 295-302 (2015-03-10)
Atherosclerosis is a chronic inflammatory disease that is initiated by the retention and accumulation of low-density lipoprotein in the artery, leading to maladaptive response of cells from the immune system and vessel wall. Strong evidence implicates indoleamine 2,3-dioxygenase (IDO), the

Protocols

Preparation from Packaged Powder, Preparation from Basal Salt Solutions, Banana Powder Preparation and Use, Coconut Water Preparation and Use

3-D Culture Matrix™ Collagen I facilitates cellular growth and differentiation in vitro, offering versatile applications in biomedical research.

3-D Culture Matrix™ Collagen I facilitates cellular growth and differentiation in vitro, offering versatile applications in biomedical research.

3-D Culture Matrix™ Collagen I facilitates cellular growth and differentiation in vitro, offering versatile applications in biomedical research.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service