Skip to Content
Merck
All Photos(4)

Key Documents

S0625

Sigma-Aldrich

D-(−)-Salicin

≥99% (GC)

Synonym(s):

2-(Hydroxymethyl)phenyl-β-D-glucopyranoside, Salicoside, Salicyl alcohol glucoside, Saligenin β-D-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O7
CAS Number:
Molecular Weight:
286.28
Beilstein:
89593
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥99% (GC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

OC[C@H]1O[C@@H](Oc2ccccc2CO)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C13H18O7/c14-5-7-3-1-2-4-8(7)19-13-12(18)11(17)10(16)9(6-15)20-13/h1-4,9-18H,5-6H2/t9-,10-,11+,12-,13-/m1/s1

InChI key

NGFMICBWJRZIBI-UJPOAAIJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Salicin is a non-phenolic glucosidic compound[1] extracted from meadowsweet (Filipendula ulmaria L). It is majorly used as a substitute for quinine. Salicin can be used as a therapeutic for patients suffering from rheumatic fever.[2] Salicin acts a metabolic precursor for salicylic acid.[3] It is a novel phytochemical that exhibits immunological cross functions in plants and humans. Salicin facilitates growth and reproduction in plants. In addition, It also protects plants against biotic and abiotic stress.[4]

Application

D-(-)-Salicin has been used:
  • to study its in vitro anticoagulant and antiplatelet activities[1]
  • as a standard in high performance liquid chromatography method (HPLC) for quantitation of salicin from willow plant[3][5]
  • as a tastant in taste threshold assay[6]
  • as a constituent of nutrient agar-salicin medium for selective isolation of Lactobacillus paracasei[7]

Biochem/physiol Actions

D-(−)-Salicin exerts anti-rheumatism and anti-inflammatory activities.[8] It inhibits lipopolysaccharide (LPS) induced inflammation in in vitro and in vivo studies.[8] It regulates different signaling pathways such as nuclear factor kappa B (NF-κB) and mitogen-activated protein kinase (MAPK).[8] It also regulates cytokines concentration such as tumor necrosis factor-alpha(TNF-α), interleukin-1β, interleukin-6, and interleukin-10.[8]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of culture media for selective enumeration of probiotic strains of lactobacilli and bifidobacteria in combination with yoghurt or cheese starters
Van de Casteele S, et al.
International dairy journal, 16(12), 1470-1476 (2006)
Autofluorescence-based identification and functional validation of antennal gustatory sensilla in a specialist leaf beetle
Pentzold S, et al.
Frontiers in Physiology, 10, 343-343 (2019)
Maik Behrens et al.
Chemical senses, 38(6), 475-484 (2013-05-02)
Mutational polymorphism in the TAS2R38 bitter taste receptor is a key determinant of threshold taste detection of isolated compounds, such as phenylthiocarbamide (PTC) and propylthiouracil (PROP), as well as complex orosensation-mediated traits such as diet choice and smoking habits. These
Drug discovery: a history (2005)
Dual anticoagulant/antiplatelet persulfated small molecules
Correia-da-Silva M, et al.
European Journal of Medicinal Chemistry, 46(6), 2347-2358 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service