Skip to Content
Merck
All Photos(7)

Key Documents

S0130

Sigma-Aldrich

Streptozocin

≥75% α-anomer basis,≥98% (HPLC), powder, DNA alkylating agent

Synonym(s):

STZ, Streptozotocine, N-(Methylnitrosocarbamoyl)-α-D-glucosamine, Streptozotocin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15N3O7
CAS Number:
Molecular Weight:
265.22
Beilstein:
2060675
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder

Assay

≥75% α-anomer basis
≥98% (HPLC)

form

powder

color

white to light yellow

mp

121 °C (dec.) (lit.)

antibiotic activity spectrum

neoplastics

Mode of action

DNA synthesis | interferes

storage temp.

−20°C

SMILES string

CN(N=O)C(=O)N[C@H]1[C@@H](O)O[C@H](CO)[C@@H](O)[C@@H]1O

InChI

1S/C8H15N3O7/c1-11(10-17)8(16)9-4-6(14)5(13)3(2-12)18-7(4)15/h3-7,12-15H,2H2,1H3,(H,9,16)/t3-,4-,5-,6-,7+/m1/s1

InChI key

ZSJLQEPLLKMAKR-GKHCUFPYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Streptozocin (STZ) is an antineoplastic antibiotic. This N-nitroso-containing compound acts as a nitric oxide donor in pancreatic islets. It is a methylnitrosourea attached to a single molecule of glucosamine and has a high affinity for the cell surface glucose transporter glucose transporter type 2 (GLUT2), which is highly expressed on beta cells of the islets of Langerhans. Streptozotocin cannot pass through the blood-brain barrier, but its metabolites are usually seen in the cerebrospinal fluid.

In cell culture medium, it has been shown to have a biological half-life of approximately 19 minutes. It is a strong methylating agent that interacts with DNA in vitro to produce methylated purines. It can impact glucose metabolism because STZ is particularly toxic to the insulin-producing beta cells of the pancreas in mammals and is also an effective antibiotic against Gram-negative bacteria. It inhibits the synthesis of DNA in microorganisms and mammalian cells by alkylation and cross-linking the strands of DNA and affects all stages of the mammalian cell cycle. STZ is considered mutagenic, carcinogenic, and possibly teratogenic in humans. It is cytotoxic to the neuroendocrine tumor cell lines that express the GLUT2 glucose transporter.

Application

Streptozocin or streptozotocin is used to induce type 1 diabetes for medical research.

Biochem/physiol Actions

An N-nitroso-containing compound that acts as a nitric oxide donor in pancreatic islets; induces death of insulin-secreting cells, producing an animal model of diabetes. Potent DNA methylating agent that induces chromosomal breakage.
An N-nitroso-containing compound that acts as a nitric oxide donor in pancreatic islets; induces death of insulin-secreting cells, producing an animal model of diabetes. Potent DNA methylating agent that induces chromosomal breakage. Cytotoxic to neuroendocrine tumor cell lines that express the GLUT2 glucose transporter.

Other Notes

Mixed anomers

also commonly purchased with this product

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Flam. Sol. 1 - Muta. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Housekeeping proteins: How useful are they in skeletal muscle
diabetes studies and muscle hypertrophy models?
Marco
Analytical Biochemistry, 504, 38-40 (2016)
Streptozocin
Papich MG
Saunders Handbook of Veterinary Drugs, 742-743 (2016)
Streptozocin
Papich MG
Journal of Separation Science, 742-743 (2016)
The Streptozotocin-Induced Diabetic Nude Mouse Model: Differences between Animals from Different Sources
Melanie L Graham
Comparative Medicine, 61(4), 356-360 (2011)
Acute stimulation of brain mu opioid receptors inhibits glucosestimulated
insulin secretion via sympathetic innervation
Eva Tuduri
Neuropharmacology, 110, 322-332 (2016)

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service