Skip to Content
Merck
All Photos(1)

Key Documents

RAB1151

Sigma-Aldrich

Rat OPN ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

species reactivity

mouse, rat

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum sample(s)
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 80 pg/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... Spp1(25353)

General description

This ELISA antibody pair detects rat and mouse OPN. Other species are not determined.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H T Nacaroglu et al.
Allergologia et immunopathologia, 45(6), 534-540 (2017-06-25)
It is thought that airway inflammation is more common in obese asthmatic patients because inflammation is harder to control and does not respond well to glucocorticoid treatment. This study's aim was to investigate the effect of obesity on airway and
Jessica F Olive et al.
PloS one, 13(6), e0198790-e0198790 (2018-06-14)
Gene editing protocols often require the use of a subcloning step to isolate successfully edited cells, the behavior of which is then compared to the aggregate parental population and/or other non-edited subclones. Here we demonstrate that the inherent functional heterogeneity
Shirley T Padilla et al.
Biomolecules, 10(11) (2020-11-05)
Acquired immunodeficiency syndrome (AIDS) complicated with tuberculosis (TB) is a global public issue. Due to the paucity of bacteria in AIDS/TB, blood-based biomarkers that reflect disease severity are desired. Plasma levels of matricellular proteins, such as osteopontin (OPN) and galectin-9
Machiko Honda et al.
Journal of cell communication and signaling, 14(2), 223-232 (2020-02-18)
Previously, we reported that an extracellular matrix protein, osteopontin (OPN), is involved in various autoimmune diseases using a neutralizing polyclonal antibody against OPN generated in rabbits. However, the antibody cannot be used for long-term mouse models of chronic inflammatory disease
S Meraviglia et al.
Oncoimmunology, 6(10), e1347742-e1347742 (2017-11-11)
γδ T cells usually infiltrate many different types of cancer, but it is unclear whether they inhibit or promote tumor progression. Moreover, properties of tumor-infiltrating γδ T cells and those in the corresponding normal tissue remain largely unknown. Here we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service