Skip to Content
Merck
All Photos(1)

Key Documents

RAB1069

Sigma-Aldrich

Human L1CAM ELISA Kit

for cell culture supernatants, plasma, and serum samples

Synonym(s):

N-CAM-L1, NCAM-L1, Neural cell adhesion molecule L1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum sample(s)
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.8 pg/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... L1CAM(3897)

General description

This ELISA antibody pair detects human L1CAM. Other species not yet determined.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seungmin Yu et al.
Journal of microbiology and biotechnology, 27(9), 1617-1627 (2017-07-29)
Dietary approaches using structured lipids, including medium-chain fatty acids and diacylglycerols, have been adopted for the prevention of obesity-induced chronic inflammation. In an extension to previous studies, medium-chain fatty acid-diacylglycerol enriched dietary oil (MCDG) was prepared by interesterification of canola
Ji Young Je et al.
European journal of pharmacology, 892, 173689-173689 (2020-11-01)
Portulaca oleracea L. is used as a folk medicine in many countries because of its wide range of pharmacological effects. HM-chromanone, isolated from P. oleracea using bioassay-guided fractionation and HPLC, belongs to the homoisoflavonoid group and has been shown to
Ling-Yu Chu et al.
Journal of Cancer, 11(18), 5395-5402 (2020-08-04)
Background: Low serum L1 cell adhesion molecule (L1CAM) has been found in several malignant tumors. Here, we aimed to evaluate the diagnostic potential for serum L1CAM in patients with gastric cancers (GC) and esophagogastric junction adenocarcinoma (EJA). Methods: Enzyme-linked immunosorbent
Hui Dai et al.
Journal of the American Heart Association, 8(3), e011155-e011155 (2019-01-29)
Background Whether chronic obstructive sleep apnea ( OSA ) could promote epicardial adipose tissue ( EAT ) secretion of profibrotic adipokines, and thereby contribute to atrial fibrosis, and the potential therapeutic effects of metoprolol remain unknown. Methods and Results A
Maria Tampaki et al.
Cancers, 12(1) (2020-01-19)
Considering the increasing importance of immune checkpoints in tumor immunity we investigated the clinical relevance of serum T-cell immunoglobulin and mucin domain-3 (TIM-3) in patients with hepatocellular carcinoma (HCC). Serum TIM-3 levels were measured and their association with HCC stage

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service