Skip to Content
Merck
All Photos(1)

Key Documents

RAB0886

Sigma-Aldrich

Rat CD40LG / CD40 Ligand ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

rat

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.45 ng/mL
standard curve range: 0.410-100 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

The antibody pair provided in this kit recognizes Rat CD40 Ligand.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Girish Viswanathan et al.
The Journal of clinical endocrinology and metabolism, 99(6), E1050-E1054 (2014-03-19)
Higher serum TSH levels, both within the reference range and in those with subclinical hypothyroidism (SCH), have been associated with increased risk of atherosclerosis and cardiovascular (CV) events in a number of cross-sectional and longitudinal studies. Our objective was to
Gage Brummer et al.
Oncogene, 39(11), 2275-2289 (2019-12-13)
Breast cancer is the second leading cause of cancer-related deaths for women, due mainly to metastatic disease. Invasive tumors exhibit aberrations in recruitment and activity of immune cells, including decreased cytotoxic T cells. Restoring the levels and activity of cytotoxic
Xingli Xu et al.
Experimental and therapeutic medicine, 19(1), 137-146 (2019-12-20)
The present study aimed to clarify the effect of bolus intracoronary nicorandil on inflammatory, oxidative and adherent indicators in patients with acute coronary syndrome (ACS) undergoing percutaneous coronary intervention (PCI). This randomized controlled trial (RCT) was performed to detect the
Leonardo Lorente et al.
Thrombosis research, 134(4), 832-836 (2014-08-16)
Serum soluble CD40 Ligand (sCD40L) levels, which exhibit prothrombotic and proinflammatory properties, have not been studied in patients with traumatic brain injury (TBI). Thus, the objective of this study was to determine whether serum sCD40L levels are associated with severity
Adem Aydın et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 32(1), 151-158 (2013-10-12)
In this study, analysis and comprehensive comparison of neurogenic differentiation capacity of human bursal tissue-derived-stem cells (hBT-SCs) was aimed with human bone marrow derived mesenchymal stem cells (hBM-MSCs). hBT-SCs was isolated from subacromial bursa tissue (n = 3) by collagen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service