Skip to Content
Merck
All Photos(1)

Documents

RAB0822

Sigma-Aldrich

Mouse Lcn2 / Neutrophil Gelatinase-associated Lipocalin ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.02 ng/mL
standard curve range: 0.024-100 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Lcn2(16819)

General description

The antibody pair provided in this kit recognizes mouse Lipocalin-2/NGAL.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Vanmassenhove et al.
International urology and nephrology, 46(10), 2003-2008 (2014-09-06)
The pathophysiology of septic acute kidney injury (AKI) is incompletely understood, and there is controversy on the role of renal hypoperfusion in early sepsis. We hypothesized that renal hypoperfusion plays a role in early sepsis and that there is a
María Teresa Torres-Salido et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 29(9), 1740-1749 (2014-04-09)
One of the challenges of treating patients with lupus nephritis (LN) is to accurately assess disease activity and predict its outcome. Since renal-biopsy cannot be performed routinely, new surrogate biomarkers are needed. We evaluated neutrophil gelatinase-associated lipocalin (NGAL), to predict
Cristiano Dias et al.
Kidney & blood pressure research, 38(2-3), 186-195 (2014-04-16)
Rosiglitazone (RGL) has been used to ameliorate lipids homeostasis and also to treat inflammatory diseases. However, RGL may reduce renal blood flow and glomerular filtration rate (GFR) predisposing to acute kidney injury (AKI). We investigated whether the treatment with RGL
Maria L Balmer et al.
Science translational medicine, 6(237), 237ra66-237ra66 (2014-05-23)
A prerequisite for establishment of mutualism between the host and the microbial community that inhabits the large intestine is the stringent mucosal compartmentalization of microorganisms. Microbe-loaded dendritic cells trafficking through lymphatics are arrested at the mesenteric lymph nodes, which constitute
Melina Vega de Ceniga et al.
Annals of vascular surgery, 28(7), 1642-1648 (2014-03-19)
Abdominal aortic aneurysms (AAAs) are currently followed with serial ultrasound or computed tomography scanning diameter measurements, but evidence shows that AAA expansion is mostly discontinuous and quite unpredictable in any given patient. A reliable predictive model of AAA growth and/or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service