RAB0748
Human TNNT1 / Troponin T, Slow Skeletal Muscle ELISA Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type serum
sample type plasma
sample type cell culture supernatant(s)
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.35 ng/mL
standard curve range: 0.391-25 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... TNNT1(7138)
General description
Troponin T1, slow skeletal type (TNNT1) is encoded by the gene mapped to human chromosome 19q13.4. The encoded protein is composed of 261 amino acids and is expressed specifically in slow skeletal muscle.
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Biochem/physiol Actions
Troponin T plays a vital role in calcium regulation of striated muscle contraction and relaxation. The alternative splicing regulation of troponin T1, slow skeletal type (TNNT1) expression has been implicated in regulation of muscle contractility in physiological and pathophysiological adaptations. Mutation in the gene is associated with the development of Nemaline myopathy.
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Bergita Ganse et al.
Experimental cell research, 399(2), 112463-112463 (2021-01-02)
Artificial gravity is a potential countermeasure to attenuate effects of weightlessness during long-term spaceflight, including losses of muscle mass and function, possibly to some extent attributable to disturbed neuromuscular interaction. The 60-day AGBRESA bed-rest study was conducted with 24 participants
Histone deacetylase inhibition elicits an evolutionarily conserved self-renewal program in embryonic stem cells.
Ware C B, et al.
Cell Stem Cell, 4(4), 359-369 (2009)
TNNT1, TNNT2, and TNNT3: Isoform Genes, Regulation, and Structure-Function Relationships.
Wei B and Jin J P
Gene, 582(1), 1-13 (2016)
Rui Tian et al.
Experimental cell research, 357(2), 328-334 (2017-06-08)
Idiopathic pulmonary fibrosis (IPF) is a chronic, progressive and irreversible lung disease. Studies have shown that epithelial-mesenchymal transition (EMT) plays an important role in the development of IPF. The NLRP3 inflammasome is reported to be activated and play an important
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service