Skip to Content
Merck
All Photos(1)

Key Documents

RAB0744

Sigma-Aldrich

Human TLR2 / Toll-like Receptor 2 ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.32 ng/mL
standard curve range: 0.328-80 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TLR2(7097)

General description

The antibody pair provided in this kit recognizes human Toll-like Receptor 2.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheng Zhang et al.
Experimental and therapeutic medicine, 17(1), 17-26 (2019-01-18)
The hypoxia-reoxygenation process of obstructive sleep apnea (OSA) may cause oxidative stress injury of the kidney, but the molecular mechanisms are not clear. The present study aimed to investigate whether high mobility group box 1 protein (HMGB1) and its subsequent
Kuo-Tung Huang et al.
PloS one, 15(2), e0228958-e0228958 (2020-02-19)
Obstructive sleep apnea (OSA) is a syndrome leading to chronic intermittent hypoxia, and the up-regulation of toll-like receptors (TLR) 2 and 6 on peripheral blood cells has been reported. We hypothesized that DNA methylation in TLR2 and TLR6 genes may
Yun Yang et al.
Journal of reproductive immunology, 142, 103192-103192 (2020-09-21)
Neisseria gonorrhoeae (N.g) is Gram-negative bacteria and can lead to endometritis in female. Toll-like receptors regulate immune response in various diseases. However, the roles of TLR2 and TLR4 in. Neisseria gonorrhoeae-induced infection damage in human endometrial epithelia were investigated. hEECs
Zdenka Holubcova et al.
Perfusion, 32(4), 269-278 (2016-11-18)
The aim was to evaluate the association between perioperative inflammatory biomarkers and atrial fibrillation (AF) in cardiac surgical patients. Forty-two patients undergoing cardiac surgery were divided into three groups according to the occurrence of AF: Group A (n = 22)
Monica Gioia Marazzi et al.
International journal of immunopathology and pharmacology, 31, 394632017749356-394632017749356 (2017-12-19)
There is still no "gold standard" for the diagnosis and prognosis of post-operative periprosthetic joint infection (PJI). Among serum biomarkers, an emerging molecule is presepsin, the soluble fraction of CD14, recently described in other settings as a powerful diagnostic tool

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service