Skip to Content
Merck
All Photos(1)

Documents

RAB0556

Sigma-Aldrich

Human VWF / von Willebrand Factor ELISA Kit

for serum, plasma, cell culture supernatants and urine

Synonym(s):

VWF ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)
sample type urine

assay range

inter-assay cv: <10%
intra-assay cv: <12%
sensitivity: 0.4 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... VWF(7450)

General description

Human VWF/von Willebrand factor ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro assay for the quantitative measurement of a target protein in biological samples, such as serum, plasma, cell culture supernatants, urine and cell and tissue lysates.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Human VWF/von Willebrand factor ELISA kit has been used for the quantification of endothelial marker-von Willebrand factor in bone marrow-derived mesenchymal stem cells (BMSC) and human umbilical vein endothelial cells (HUVEC).

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roland von Känel et al.
The journals of gerontology. Series A, Biological sciences and medical sciences, 75(11), 2215-2223 (2020-04-04)
This study examined whether biological mechanisms linking dementia caregiving with an increased risk of coronary heart disease can be modified by psychosocial interventions and which caregivers might benefit the most from an intervention. Spousal dementia caregivers were randomized to 12-week
J W R Sins et al.
Journal of thrombosis and haemostasis : JTH, 15(7), 1392-1402 (2017-05-01)
Essentials The role of von Willebrand Factor (VWF) in the pathophysiology of sickle cell disease is unclear. We assessed markers of VWF during admission for vaso-occlusive crisis (VOC) and steady state. VWF reactivity was higher during VOC and was associated
M M Mebius et al.
Journal of helminthology, 94, e84-e84 (2019-09-11)
In many tropical areas schistosomiasis is a major health problem causing hepatosplenic, intestinal or urogenital complaints. Hepatosplenic schistosomiasis mansoni is also characterized by blood coagulation abnormalities. Liver pathology plays a role in the development of haemostatic changes and the parasitic
Marko Stojanović et al.
Archives of physiology and biochemistry, 125(1), 44-55 (2018-02-13)
We have performed an in vitro study on isolated intact or denuded femoral artery (FA) of healthy, diabetic, and/or rats submitted to the FA occlusion. The aim was to determine the contribution of endothelium and endothelial dysfunction (ED) on serotonin-induced
Camila M de Andrade et al.
International journal of biological macromolecules, 134, 653-659 (2019-05-10)
Endothelium plays an important modulatory role due to the synthesis and secretion of molecules that act on hemostasis and fibrinolysis. As there are no literature data about the effect of crotoxin (CTX) - the main component of Crotalus durissus terrificus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service