Skip to Content
Merck
All Photos(1)

Documents

RAB0062

Sigma-Aldrich

Human CCL21 / 6ckine ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)
sample type urine

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 30 pg/mL
standard curve range: 32.76-8000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL21(6366)

General description

The Human 6Ckine ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human 6Ckine in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human 6ckine

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADCELISA 1X Assay/Sample Diluent Buffer C (Item L)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Qian et al.
Veterinary microbiology, 203, 158-166 (2017-06-18)
Newcastle disease virus-like particles (NDV VLPs) are a potential candidate vaccine, as shown by eliciting specific immune response against NDV in mice and chickens. Activation of dendritic cells (DCs) is critical to initiate immune response. However, the mechanism of how
Attilio Pingitore et al.
Molecular and cellular endocrinology, 436, 23-32 (2016-07-18)
Nerve growth factor (NGF) is a protein required for neuronal development that also has regulatory functions in non-neuronal cells. Both NGF and its membrane receptors trkA and p75(NTR) are expressed by islet β-cells. In this study we dynamically profiled NGF
Brittany J Poelaert et al.
Journal of controlled release : official journal of the Controlled Release Society, 327, 266-283 (2020-07-28)
Neuroblastoma is the most commonly diagnosed extracranial solid tumor in children. The patients with aggressive metastatic disease or refractory/relapsed neuroblastoma currently face a dismally low chance of survival. Thus, there is a great need for more effective therapies for this
Xiaofeng Guo et al.
Scientific reports, 10(1), 14386-14386 (2020-09-03)
Regulatory CD19+CD24hiCD27+ B cells were proved to be numerically decreased and functionally impaired in the peripheral blood (PB) from rheumatoid arthritis (RA), with the potential of converting into osteoclast-priming cells. However, the distribution and function of CD19+CD24hiCD27+ B cells in
G Shilpa et al.
Phytotherapy research : PTR, 31(12), 1892-1902 (2017-10-19)
Epoxyazadiradione (EAD) is an important limonoid present in Neem (Azadirachta indica) plant. In the present study, we have purified EAD from Neem seed and studied its anticancer potential in human cervical cancer (HeLa) cells. Cell proliferation inhibition studies indicated that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service