Skip to Content
Merck
All Photos(2)

Key Documents

RAB0042

Sigma-Aldrich

Human Eotaxin / CCL11 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type urine
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 45 pg/mL
standard curve range: 49.15-12000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL11(6356)

General description

The Human Eotaxin ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human Eotaxin in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human Eotaxin

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Nair et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 44(9), 1200-1203 (2014-08-19)
Inhaled cationic airway lining modulator (iCALM) is a cationic aerosol therapy comprised of 1.29% calcium chloride dissolved in 0.9% isotonic saline that enhances the biophysical barrier function of the airway lining fluid and primes the host defense response. It's ability
Xiao Na Ge et al.
Glycobiology, 24(8), 715-727 (2014-05-06)
Heparan sulfate (HS) proteoglycans (HSPGs) participate in several aspects of inflammation because of their ability to bind to growth factors, chemokines, interleukins and extracellular matrix proteins as well as promote inflammatory cell trafficking and migration. We investigated whether HSPGs play
Chan-Mi Jeon et al.
International immunopharmacology, 22(2), 414-419 (2014-07-30)
Siegesbeckia glabrescens (SG) is a plant growing in Korea that is used as a traditional medicine for various inflammatory diseases. In this study, we investigated the protective effects of SG extract on allergic asthma in an ovalbumin (OVA)-induced asthma murine
Nikola Sladojevic et al.
Neurobiology of disease, 67, 57-70 (2014-03-25)
Proinflammatory mediators trigger intensive postischemic inflammatory remodeling of the blood-brain barrier (BBB) including extensive brain endothelial cell surface and junctional complex changes. Junctional adhesion molecule-A (JAM-A) is a component of the brain endothelial junctional complex with dual roles: paracellular route

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service