RAB0039
Cholecystokinin EIA Kit
for serum, plasma, culture supernatant and cell lysates
Synonym(s):
CCK
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41123300
NACRES:
NA.32
Recommended Products
species reactivity
rat, mouse, human
technique(s)
ELISA: suitable
enzyme immunoassay: suitable
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... CCK(885)
General description
Cholecystokinin (CCK) belongs to the gastrointestinal hormone family. It is a neuropeptide hormone and neurotransmitter usually present in gastrointestinal (GI) tract and the central nervous system (CNS). It is produced by neurons, and endocrine cells.
Immunogen
Cholecystokinin (CCK), synthetic peptide
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Biochem/physiol Actions
Cholecystokinin (CCK) functions include, trophic effects on pancreas, gall bladder contraction, intestinal motility, and bowel movements. It has roles in various neuropsychiatric and neurological disorders. CCK is satiety peptide and regulates apettite for a short duration. Overproduction of CCK is prominent in metastatic endocrine tumors, and hence it is recognize as a diagnostic marker.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Cholecystokinin: A multi-functional molecular switch of neuronal circuits
Lee SY and Soltesz I
Developmental neurobiology, 71(1), 83-91 (2011)
Regulation of Gastrointestinal Mucosal Growth (2010)
Postprandial profiles of CCK after high fat and high carbohydrate meals and the relationship to satiety in humans
Gibbons C, et al.
Peptides, 77, 3-8 (2016)
Cholecystokinin in plasma predicts cardiovascular mortality in elderly females
Goetze JP, et al.
International Journal of Cardiology, 209, 37-41 (2016)
L I Larsson et al.
Brain research, 165(2), 201-218 (1979-04-13)
Immunocytochemistry and radioimmunochemistry demonstrate the occurrence of the gastrointestinal hormone cholecystokinin (CCK) in both the central and peripheral nervous system of the guinea pig. CCK nerves are particularly numerous in the neocortex, the hippocampus, the amygdaloid nuclei, the hypothalamus, the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service