Skip to Content
Merck
All Photos(4)

Documents

R5500

Sigma-Aldrich

Ribonuclease A from bovine pancreas

Type XII-A, ≥90% (SDS-PAGE), 75-125 Kunitz units/mg protein

Synonym(s):

Pancreatic Ribonuclease, RNAsea, RNase A, Ribonucleate 3′-pyrimidinooligonucleotidohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bovine pancreas

type

Type XII-A

Assay

≥90% (SDS-PAGE)

form

lyophilized powder

specific activity

75-125 Kunitz units/mg protein

mol wt

~13,700

technique(s)

cell based assay: suitable

impurities

salt, essentially free

suitability

suitable for mRNA or total RNA extracted from cells and tissues

application(s)

diagnostic assay manufacturing

foreign activity

protease, essentially free

storage temp.

−20°C

InChI

1S/C9H14N4O3/c10-2-1-8(14)13-7(9(15)16)3-6-4-11-5-12-6/h4-5,7H,1-3,10H2,(H,11,12)(H,13,14)(H,15,16)

InChI key

CQOVPNPJLQNMDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

RNase A, Ribonuclease A, is an endoribonuclease that cleaves the phosphodiester bonds of single strand RNA after pyrimidine nucleotides. It attacks at the 3′ phosphate end (For example pG-pG-pC-pA-pG will be cleaved to give pG-pG-pCp and A-pG). The highest activity is exhibited with single stranded RNA. RNase A is a single chain polypeptide containing 4 disulfide bridges. In contrast to RNase B, it is not a glycoprotein. Ribonucleases do not hydrolyze DNA, because the DNA lacks 2′-OH groups essential for the formation of cyclic intermediates. RNase A can also hydrolyze RNA from protein samples. RNase A can be inhibited by alkylation of His12 and His119 and activated by potassium and sodium salts. RNAse is inhibited in the presence of heavy metal ions. RNase is also inhibited competitively by DNA.

Application

  • RNase A is used to remove RNA from DNA plasmid and genomic DNA preparations and protein samples.
  • RNase A is also used in RNA sequence analysis and protection assays.
  • RNase A has been used as a tool for computer-aided drug design.
  • RNase A supports the analysis of RNA sequences.
  • RNase A hydrolyze RNA contained in protein samples.
  • Purification of DNA is supported by RNase A.

Biochem/physiol Actions

Ribonuclease A is an endoribonuclease that cleaves single stranded RNA after pyrimidine nucleotides. It attacks at the 3′ phosphate end. Ribonucleases do not hydrolyze DNA, because the DNA lacks 2′-OH groups essential for the formation of cyclic intermediates. RNase can also hydrolyze RNA from protein samples. RNase A can be inhibited by alkylation of His12 and His119 and activated by potassium and sodium salts.

Features and Benefits

Our highly stable Ribonuclease A, RNase A, is suitable for removal of RNA, RNA sequencing, and DNA purification.

Preparation Note

Salt fractionated and chromatographically purified.

Analysis Note

Protein determined by E.

inhibitor

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amy B Emerman et al.
Methods in molecular biology (Clifton, N.J.), 1413, 303-324 (2016-05-20)
RNAs associate with the mitotic spindle in a variety of organisms, where they can spatially regulate protein production, ensure their proper segregation during cell division, or perform translation-independent roles in spindle formation. The identification of spindle-associated RNAs is an important
Vlad Zabrouskov et al.
Biochemistry, 45(3), 987-992 (2006-01-18)
Although deamidation at asparagine and glutamine has been found in numerous studies of a variety of proteins, in almost all cases the analytical methodology that was used could detect only a single site of deamidation. For the extensively studied case
Amaya Albalat et al.
Methods in molecular biology (Clifton, N.J.), 984, 153-165 (2013-02-07)
The analysis of proteins and peptides in biological fluids is becoming more important as they are potential sources of diagnostic biomarkers of disease. The complexity of body fluids is such that no single technique can both identify and quantify all
Xin-Miao Fu et al.
Biochimica et biophysica acta, 1814(4), 487-495 (2011-01-18)
Protein disulfide isomerase (PDI) and its pancreatic homolog (PDIp) are folding catalysts for the formation, reduction, and/or isomerization of disulfide bonds in substrate proteins. However, the question as to whether PDI and PDIp can directly attack the native disulfide bonds
Aarón Millán-Oropeza et al.
Proteomes, 10(1) (2022-01-26)
In proteomics, it is essential to quantify proteins in absolute terms if we wish to compare results among studies and integrate high-throughput biological data into genome-scale metabolic models. While labeling target peptides with stable isotopes allow protein abundance to be

Protocols

This procedure may be used for determination of Ribonuclease A (RNase A) activity.

This procedure may be used for determination of Ribonuclease A (RNase A) activity.

This procedure may be used for determination of Ribonuclease A (RNase A) activity.

This procedure may be used for determination of Ribonuclease A (RNase A) activity.

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service