Skip to Content
Merck
All Photos(1)

Documents

R4877

Sigma-Aldrich

Rennin from calf stomach

≥20 units/mg protein

Synonym(s):

Chymosin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥20 units/mg protein

purified by

crystallization

composition

Protein, ≥40%

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Rennin, also known as chymosin, is a milkclotting acid proteinase produced in the stomach of a calf. It is used in cheesemaking and to study neonatal gastric digestion .

Biochem/physiol Actions

Rennin, a 323 amino acid chain, is secreted as an inactive precursor which is then converted into an active enzyme through limited proteolysis. It cleaves the peptide bond between phenylalanine and methionine in K-Casein.

Packaging

Package size based on protein content

Unit Definition

One unit will coagulate 10 mL of milk per min at 30 °C.

Physical form

Lyophilized powder containing sodium chloride

Analysis Note

Protein determined by biuret.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kirsten Kastberg Møller et al.
Journal of agricultural and food chemistry, 60(21), 5454-5460 (2012-05-09)
Bovine chymosin constitutes a traditional ingredient for enzymatic milk coagulation in cheese making, providing a strong clotting capacity and low general proteolytic activity. Recently, these properties were surpassed by camel chymosin, but the mechanistic difference behind their action is not
Neil D Rawlings et al.
Nucleic acids research, 40(Database issue), D343-D350 (2011-11-17)
Peptidases, their substrates and inhibitors are of great relevance to biology, medicine and biotechnology. The MEROPS database (http://merops.sanger.ac.uk) aims to fulfil the need for an integrated source of information about these. The database has hierarchical classifications in which homologous sets
Xin-ping Li et al.
Xi bao yu fen zi mian yi xue za zhi = Chinese journal of cellular and molecular immunology, 28(7), 715-717 (2012-07-10)
To optimize the prochymosin (pCHY) gene codons and express the gene in Escherichia coli (E.coli), and to prepare its antiserum and detect chymosin protein specifically. According to codon usage bias of E.coli, prochymosin gene sequence was synthesized based on the
Jung-Feng Hsieh et al.
Journal of agricultural and food chemistry, 60(8), 2039-2045 (2012-02-07)
Chymosin-induced coagulation of individual milk proteins during incubation at 30 °C was investigated using a proteomic approach. The addition of chymosin (0.006 units/mL) caused the milk proteins to coagulate after a 3 h incubation period. Approximately 88% of the milk
X P Jiang et al.
World journal of microbiology & biotechnology, 28(5), 2087-2093 (2012-07-19)
Chymosin can specifically break down the Phe105-Met106 peptide bond of milk κ-casein to form insoluble para-κ-casein, resulting in milk coagulation, a process that is used in making cheese. In this study, in order to obtain an alternative milk coagulant which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service