Skip to Content
Merck
All Photos(1)

Key Documents

R3530

Sigma-Aldrich

Rifabutin

>98% (HPLC), powder, antibiotic

Synonym(s):

Ansamycin, Ansatipine (Farmitalia), LM-427, Mycobutin (Farmitalia)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H62N4O11
CAS Number:
Molecular Weight:
847.00
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Rifabutin, >98% (HPLC), powder

Quality Level

Assay

>98% (HPLC)

form

powder

storage condition

protect from light

solubility

DMSO: >5 mg/mL

antibiotic activity spectrum

neoplastics

Mode of action

enzyme | inhibits

originator

Johnson & Johnson

shipped in

wet ice

storage temp.

−20°C

SMILES string

[N+H]1(CCC2(NC3=C4NC(=O)\C(=C/C=C/[C@@H]([C@@H]([C@H]([C@H]([C@H]([C@@H]([C@@H]([C@H](\C=C\O[C@]5(Oc6c(c(c(c(c6C)[O-])C4=O)C3=N2)C5=O)C)OC)C)OC(=O)C)C)O)C)O)C)\C)CC1)CC(C)C

InChI

1S/C46H62N4O11/c1-22(2)21-50-18-16-46(17-19-50)48-34-31-32-39(54)28(8)42-33(31)43(56)45(10,61-42)59-20-15-30(58-11)25(5)41(60-29(9)51)27(7)38(53)26(6)37(52)23(3)13-12-14-24(4)44(57)47-36(40(32)55)35(34)49-46/h12-15,20,22-23,25-27,30,37-38,41,49,52-54H,16-19,21H2,1-11H3,(H,47,57)/b13-12+,20-15+,24-14-/t23-,25+,26+,27+,30-,37-,38+,41+,45-/m0/s1

InChI key

ATEBXHFBFRCZMA-VXTBVIBXSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Rifabutin is an antibiotic; antitumor.
Rifabutin is an antibiotic; antitumor. Rifabutin interferes with HSP-90 molecular chaperone, enhances ubiquitination and protein degradation, and inactivates bacterial RNA polymerase.

Features and Benefits

This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marina Pinheiro et al.
Journal of medicinal chemistry, 56(2), 417-426 (2012-12-12)
This work focuses on the interaction of rifabutin (RFB), a naphthalenic ansamycin, with membrane models. Since the therapeutic and toxic effects of this class of drugs are strongly influenced by their lipid affinity, we concerned specifically on the ability of
Frederick A Sirgel et al.
PloS one, 8(3), e59414-e59414 (2013-03-26)
Genetically related Mycobacterium tuberculosis strains with alterations at codon 516 in the rpoB gene were observed amongst a substantial number of patients with drug resistant tuberculosis in the Eastern Cape Province (ECP) of South Africa. Mutations at codon 516 are
C Y Tay et al.
Alimentary pharmacology & therapeutics, 36(11-12), 1076-1083 (2012-10-18)
Helicobacter pylori eradication rates with standard triple therapy are declining worldwide. The optimal management of H. pylori is evolving and new treatment combinations for antibiotic resistant H. pylori strains are required, especially for patients with penicillin allergy. To review the
Matthew D Hickey et al.
Liver transplantation : official publication of the American Association for the Study of Liver Diseases and the International Liver Transplantation Society, 19(4), 457-461 (2013-03-26)
Latent tuberculosis infection is an important problem for solid organ transplant recipients because of the frequency of its occurrence and its potential for reactivation. Because of the high mortality rate associated with active tuberculosis infections in transplant recipients, guidelines from
Jakko van Ingen et al.
American journal of respiratory and critical care medicine, 186(6), 559-565 (2012-06-30)
Currently recommended multidrug treatment regimens for Mycobacterium avium complex (MAC) lung disease yield limited cure rates. This results, in part, from incomplete understanding of the pharmacokinetics and pharmacodynamics of the drugs. To study pharmacokinetics, pharmacodynamics, and drug interactions of multidrug

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service