Skip to Content
Merck
All Photos(1)

Key Documents

R1903

Sigma-Aldrich

RS 102895 hydrochloride

≥98% (HPLC), powder

Synonym(s):

1′-[2-[4-(Trifluoromethyl)phenyl]ethyl]-spiro[4H-3,1-benzoxazine-4,4′-peperidin]-2(1H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21F3N2O2 · HCl
Molecular Weight:
426.86
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥98% (HPLC)

form

powder

mol wt

426.86 g/mol

packaging

pkg of 5 mg

storage condition

desiccated

color

white

solubility

DMSO: >10 mg/mL

storage temp.

20-25°C

SMILES string

Cl.FC(F)(F)c1ccc(CCN2CCC3(CC2)OC(=O)Nc4ccccc34)cc1

InChI

1S/C21H21F3N2O2.ClH/c22-21(23,24)16-7-5-15(6-8-16)9-12-26-13-10-20(11-14-26)17-3-1-2-4-18(17)25-19(27)28-20;/h1-8H,9-14H2,(H,25,27);1H

InChI key

KRRISOFSWVKYBF-UHFFFAOYSA-N

General description

RS 102895 hydrochloride is a small molecule antagonist of C-C motif chemokine receptor 2 (CCR2) which belongs to the spiropiperidine (SP) class.

Application

RS 102895 hydrochloride has been used in bromodeoxyuridine (5-bromo-2′-deoxyuridine, BrdU) cell proliferation assay to study effect of IL33 (interleukin 33) on decidual stromal cell proliferation in present of the antagonist and to study the effects of antagonists on the accumulation of bone marrow-derived microglia.

Biochem/physiol Actions

RS 102895 hydrochloride has a high affinity to the β subunit of the C-C motif chemokine receptor 2 (CCR2) and binds specifically to it. This results in the inhibition of CCR2 signaling. It also suppresses monocyte recruitment. Hence, RS 102895 may be useful for obstructing the immune suppressive effects of monocytes during early vaccine responses.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suppression of bone marrow-derived microglia in the amygdala improves anxiety-like behavior induced by chronic partial sciatic nerve ligation in mice.
Sawada A, Niiyama Y, Ataka K, et al.
Pain, 155(9), 1762-1772 (2014)
Identification of the binding site for a novel class of CCR2b chemokine receptor antagonists: binding to a common chemokine receptor motif within the helical bundle.
Mirzadegan T
The Journal of Biological Chemistry, 275(33), 25562-25571 (2000)
Bone marrow-derived microglia infiltrate into the paraventricular nucleus of chronic psychological stress-loaded mice.
Ataka K
PLoS ONE, 8(11), e81744-e81744 (2013)
Optimized dosing of a CCR2 antagonist for amplification of vaccine immunity.
Mitchell LA
International Immunopharmacology, 15(2), 357-363 (2013)
IL-33 enhances proliferation and invasiveness of decidual stromal cells by up-regulation of CCL2/CCR2 via NF-?B and ERK1/2 signaling.
Hu WT
Molecular Human Reproduction, 20(4), 358-372 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service