Skip to Content
Merck
All Photos(1)

Documents

R121

Sigma-Aldrich

S(−)-Raclopride (+)-tartrate salt

>97%, solid

Synonym(s):

3,5-Dichloro-N-(1-ethylpyrrolidin-2-ylmethyl)-2-hydroxy-6-methoxybenzamide (+)-tartrate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20Cl2N2O3 · C4H6O6
CAS Number:
Molecular Weight:
497.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

>97%

form

solid

color

white

solubility

H2O: 89 mg/mL

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O.CCN1CCC[C@H]1CNC(=O)c2c(O)c(Cl)cc(Cl)c2OC

InChI

1S/C15H20Cl2N2O3.C4H6O6/c1-3-19-6-4-5-9(19)8-18-15(21)12-13(20)10(16)7-11(17)14(12)22-2;5-1(3(7)8)2(6)4(9)10/h7,9,20H,3-6,8H2,1-2H3,(H,18,21);1-2,5-6H,(H,7,8)(H,9,10)/t9-;1-,2-/m01/s1

InChI key

QULBVRZTKPQGCR-NDAAPVSOSA-N

Gene Information

human ... DRD2(1813)

Application

S(−)-Raclopride (+)-tartrate salt has been used as a D2 receptor antagonist:
  • to study the role of the D2 receptor in mediating ventrolateral PAG (vlPAG)/dorsal raphe dopaminergic antinociceptive effect in animals
  • in pharmacological and behavioral studies performed with Thap1 null mice
  • to determine the relationship between Ca2+ transients and dopamine neuron activity in rats

Biochem/physiol Actions

Raclopride is a benzamide derivative and a selective D2 dopamine receptor antagonist.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Caution

Photosensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Lublin et al.
Psychopharmacology, 112(2-3), 389-397 (1993-01-01)
The effects of chronic treatment with dopamine (DA) D1 and D2 receptor antagonists were evaluated in eight cebus apella monkeys with mild oral dyskinesia after previous haloperidol treatment. SCH 23390 (D1 antagonist) was given daily to investigate the direct behavioural
P Protais et al.
European journal of pharmacology, 271(1), 167-177 (1994-12-12)
S 14506 (1-[-(4-fluorobenzoylamino)ethyl]-4-(7-methoxynaphthyl)piper azine hydrochloride), 8-OH-DPAT ((+/-)-8-hydroxydipropylaminotetralin hydrobromide), clozapine and raclopride were compared in some behavioural models able to characterize dopamine antagonist properties. In mice treated with apomorphine (0.75 mg/kg, s.c.), stereotyped climbing and sniffing were dose dependently antagonized by
J Kopp et al.
Neuroscience, 47(4), 771-779 (1992-01-01)
Prolonged treatment with dopamine D2 receptor antagonists is known to elevate the density of dopamine D2 receptor binding sites in caudate-putamen and nucleus accumbens in rat and human brain. In this study we used the dopamine D2 receptor antagonist raclopride
Angel Toval et al.
Molecular neurobiology, 58(4), 1782-1791 (2021-01-05)
Improving exercise capacity during adolescence impacts positively on cognitive and motor functions. However, the neural mechanisms contributing to enhance physical performance during this sensitive period remain poorly understood. Such knowledge could help to optimize exercise programs and promote a healthy
Vaibhav R Konanur et al.
Physiology & behavior, 215, 112771-112771 (2019-12-11)
Phasic dopamine activity is evoked by reliable predictors of food reward and plays a role in cue-triggered, goal-directed behavior. While this important signal is modulated by physiological state (e.g. hunger, satiety), the mechanisms by which physiological state is integrated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service