Q50120
QAE Sephadex® A-50 chloride form
Synonym(s):
Diethyl-(2-hydroxypropyl)aminoethyl Sephadex®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
bead size
40-125 μm (dry)
pore size
~200,000 Da exclusion limit
operating pH
2-10
capacity
2.6-3.4 meq/g ion exchange capacity
compatibility
mode of use strong anion exchange chromatography
General description
Q50120-100G′s updated product number is GE17-0200-01
Application
QAE Sephadex® is used in protein chromatography, ion exchange chromatography, anion exchange media, resins and separation media. QAE Sephadex® has been used to develop methods to detect copper in water samples, resveratrol in beer, and indomethacin in both pharmaceuticals and urine samples.
Legal Information
Sephadex is a registered trademark of Cytiva
replaced by
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Purification and properties of protein methylase II.
Archives of biochemistry and biophysics, 157(2), 476-484 (1973-08-01)
Blood, 78(2), 357-363 (1991-07-15)
Activated protein C is a plasma anticoagulant. For activated protein C to function as an anticoagulant, it must form a complex with protein S. Protein S anticoagulant activity is neutralized by formation of a reversible complex with C4b binding protein
Characterization of an Apple Polygalacturonase-Inhibiting Protein (PGIP) That Specifically Inhibits an Endopolygalacturonase (PG) Purified from Apple Fruits Infected with <I>Botryosphaeria dothidea</I>.
Journal of microbiology and biotechnology, 16(8), 1192-1200 (2006)
Biochemistry, 27(16), 5939-5947 (1988-08-09)
Triosephosphate isomerase from bakers' yeast, expressed in Escherichia coli strain DF502(p12), has been purified to homogeneity. The kinetics of the reaction in each direction have been determined at pH 7.5 and 30 degrees C. Deuterium substitution at the C-2 position
Clinical chemistry, 35(1), 69-73 (1989-01-01)
The bacterial thrombolytic agent streptokinase binds to human, porcine, and chicken lactate dehydrogenase (EC 1.1.1.27; LD) isoenzyme subunit M, but not to the H or C subunits. There is amino acid sequence homology between LD and the streptokinase binding site
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service