Skip to Content
Merck
All Photos(1)

Key Documents

PZ0383

Sigma-Aldrich

Palbociclib

≥98% (HPLC), powder, CDK 4 & 6 inhibitor

Synonym(s):

6-Acetyl-8-cyclopentyl-5-methyl-2-[[5-(1-piperazinyl)-2-pyridinyl]amino]pyrido[2,3-d]pyrimidin-7(8H)-one, Ibrance, PD 0332991, PD 332991, PD-0332991, PD0332991, PD991, PF 00080665, PF-00080665, PF00080665

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H29N7O2
CAS Number:
Molecular Weight:
447.53
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Palbociclib, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

0.1 M HCl: 2 mg/mL, clear

storage temp.

room temp

SMILES string

CC(C1=C(N2C3CCCC3)N=C(NC4=CC=C(N5CCNCC5)C=N4)N=C1)=C(C(C)=O)C2=O

InChI

1S/C24H29N7O2/c1-15-19-14-27-24(28-20-8-7-18(13-26-20)30-11-9-25-10-12-30)29-22(19)31(17-5-3-4-6-17)23(33)21(15)16(2)32/h7-8,13-14,17,25H,3-6,9-12H2,1-2H3,(H,26,27,28,29)

InChI key

AHJRHEGDXFFMBM-UHFFFAOYSA-N

Biochem/physiol Actions

PF-00080665 (Palbociclib; PD 0332991) is an orally active and highly specific inhibitor against cyclin-dependent kinase 4 & 6 (IC50 = 9, 11, 15 nM, respectively, using CDK4/cycD3, CDK4/cycD1, CDK6/cycD2; IC50 >10 μM against 36 other kinases) that potently suppresses Cdk4/6-dependent cellular Rb phosphorylation (IC50 = 66 nM/pSer780 & 63 nM/pSer795; MDA-MB-435). PF-00080665 exhibits selective antiproliferation activity against Rb-positive human breast/colon/lung/leukemia cancer cultures (IC50 = 40-400 nM; IC50 >3 μM/Rb-negative MDA-MB-468 & H2009) and displays in vivo efficacy against various advanced stage human tumor xenografts in mice (12.5-150 mg/kg/day p.o.).

Storage and Stability

This chemical can be stored in solution at -80°C for 6 months and at -20°C for 1 month (protect from light).

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Muta. 2 - Repr. 2 - STOT RE 2 Oral

Target Organs

Bone marrow,male reproductive organs

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryohei Kozaki et al.
Cancers, 10(4) (2018-04-25)
Bruton's tyrosine kinase (BTK) is a key regulator of the B-cell receptor signaling pathway, and aberrant B-cell receptor (BCR) signaling has been implicated in the survival of malignant B-cells. However, responses of the diffuse large B-cell lymphoma (DLBCL) to inhibitors
Fiona J Ruiz et al.
Cancers, 13(18) (2021-09-29)
Cervical cancer tumors with undetectable HPV (HPVU) have been underappreciated in clinical decision making. In this study, two independent CC datasets were used to characterize the largest cohort of HPVU tumors to date (HPVU = 35, HPV+ = 430). Genomic
Xiaoyang Li et al.
Cell death & disease, 9(5), 446-446 (2018-04-20)
Synovial sarcoma is a highly aggressive but rare form of soft tissue malignancy that primarily affects the extremities of the arms or legs, for which current chemotherapeutic agents have not been proven to be very effective. The cyclin-dependent kinase 4/6-retinoblastoma
Shinya Kitamura et al.
Cancer science, 113(2), 802-807 (2021-12-07)
Extramammary Paget's disease (EMPD) is a rare adnexal neoplasm commonly seen in the genital areas among the senior population. The prognosis of advanced EMPD is not favorable; thus, the development of potential treatments has long been sought. Cyclin-dependent kinase (CDK)
ER
Angel L Guerrero-Zotano et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 24(11), 2517-2529 (2018-03-28)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service