Skip to Content
Merck
All Photos(3)

Key Documents

PRS3613

Sigma-Aldrich

Anti-Beclin-1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Beclin 1, autophagy related, Anti-Coiled-coil myosin-like Bcl-2-interacting protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BECN1(8678)

Related Categories

Immunogen

a 17 amino acid peptide from near the amino-terminus of human Beclin-1.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3613.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Sykora et al.
Nucleic acids research, 43(2), 943-959 (2015-01-02)
We explore the role of DNA damage processing in the progression of cognitive decline by creating a new mouse model. The new model is a cross of a common Alzheimer's disease (AD) mouse (3xTgAD), with a mouse that is heterozygous
Samar R Saleh et al.
Antioxidants (Basel, Switzerland), 11(7) (2022-07-28)
Long-term glucocorticoids can alter sperm motility, vitality, or morphology, disrupting male reproductive function. This study scrutinized the synergistic benefits of two Egyptian plants against dexamethasone (Dexa)-induced testicular and autophagy dysfunction in male rats. Phytochemical ingredients and the combination index were
Collin J Fitzpatrick et al.
Frontiers in cellular and infection microbiology, 12, 798978-798978 (2022-04-26)
Junín virus (JUNV), a New World arenavirus, is a rodent-borne virus and the causative agent of Argentine hemorrhagic fever. Humans become infected through exposure to rodent host secreta and excreta and the resulting infection can lead to an acute inflammatory
Hua Tian et al.
Experimental biology and medicine (Maywood, N.J.), 246(24), 2595-2609 (2021-09-17)
The present study aimed to investigate the role of D4F, an apolipoprotein A-I mimetic peptide, in macrophage apoptosis induced by the glycated high-density lipoprotein (gly-HDL)-induced endoplasmic reticulum (ER) stress C/EBP homologous protein (CHOP) pathway, and unravel the regulatory role of
Cynthia de la Fuente et al.
PloS one, 13(2), e0191983-e0191983 (2018-02-07)
Rift Valley fever virus (RVFV) infects both ruminants and humans leading to a wide variance of pathologies dependent on host background and age. Utilizing a targeted reverse phase protein array (RPPA) to define changes in signaling cascades after in vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service