Skip to Content
Merck
All Photos(1)

Key Documents

PROTSIL1

Sigma-Aldrich

ProteoSilver Silver Stain Kit

Synonym(s):

Silver Staining Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105322
NACRES:
NA.32

usage

 kit sufficient for 25 mini-gels (10 × 10 cm)

technique(s)

protein staining: suitable

General description

Conveniently packaged, extremely stable, and highly sensitive, ProteoSilver is an ideal product for any protein scientist. The kit contains prepared solutions of silver staining reagents along with detailed instructions to achieve optimal results. With a detection limit of 0.1 ng/mm2 of protein (BSA) and an extremely low background, ProteoSilver leads to superior detection of low abundance proteins.

Application

ProteoSilver Silver Stain Kit has been used for staining of the gels for visualizing proteins.

Features and Benefits

  • Premixed and preweighed solutions reduce time and cost of purchasing and preparing individual components
  • Optimized protocols help establish conditions for best results
  • High sensitivity and low background ensure very low abundance proteins can be detected and resolved from other proteins
  • MALDI Compatible - Protein spots of interest can be further characterized by mass spectrometry
  • Room Temperature Stability allows easy and convenient storage

Legal Information

ProteoSilver is a trademark of Sigma-Aldrich Co. LLC

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Muta. 2 - Repr. 1B - Skin Corr. 1A - Skin Sens. 1 - STOT SE 2 - STOT SE 3

Target Organs

Eyes,Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean-Paul Lasserre et al.
Electrophoresis, 27(16), 3306-3321 (2006-07-22)
Study of the complexome - all the protein complexes of the cell - is essential for a better understanding and more global vision of cell function. Using two-dimensional blue native/SDS-PAGE (2-D BN/SDS-PAGE) technology, the cytosolic and membrane protein complexes of
Yufan Zhang et al.
Materials today. Bio, 15, 100317-100317 (2022-06-28)
Polymer dots (Pdots) have been applied to imaging lymph nodes (LNs) and lymphatic vessels (LVs) in living mice and rats. However, the mechanism of absorption, distribution, metabolism, and excretion of Pdots in LNs and LVs is still unclear. Therefore, the
Yong-Feng Han et al.
The Plant cell, 28(5), 1215-1229 (2016-04-27)
The mechanism by which MORPHEUS' MOLECULE1 (MOM1) contributes to transcriptional gene silencing has remained elusive since the gene was first identified and characterized. Here, we report that two Arabidopsis thaliana PIAS (PROTEIN INHIBITOR OF ACTIVATED STAT)-type SUMO E3 ligase-like proteins
Andrea Miccoli et al.
International journal of molecular sciences, 22(7) (2021-04-04)
In jawed vertebrates, adaptive immune responses are enabled by T cells. Two lineages were characterized based on their T cell receptor (TcR) heterodimers, namely αβ or γδ peptide chains, which display an Ig domain-type sequence that is somatically rearranged. γδ
Zongyu Gao et al.
Nature communications, 13(1), 716-716 (2022-02-09)
Mitogen-activated protein kinase (MAPK) cascades play an important role in innate immunity against various pathogens in plants and animals. However, we know very little about the importance of MAPK cascades in plant defense against viral pathogens. Here, we used a

Articles

Sigma offers EZBlue™ and ProteoSilver™ reagents for protein visualization, suitable for proteomics and traditional PAGE formats.

Sigma offers EZBlue™ and ProteoSilver™ reagents for protein visualization, suitable for proteomics and traditional PAGE formats.

Sigma offers EZBlue™ and ProteoSilver™ reagents for protein visualization, suitable for proteomics and traditional PAGE formats.

Sigma offers EZBlue™ and ProteoSilver™ reagents for protein visualization, suitable for proteomics and traditional PAGE formats.

See All

Protocols

Protocol for sample preparation for cell lysis and efficient protein extraction from cultured tissues and cells for subsequent Western blotting.

Protocol for sample preparation for cell lysis and efficient protein extraction from cultured tissues and cells for subsequent Western blotting.

Protocol for sample preparation for cell lysis and efficient protein extraction from cultured tissues and cells for subsequent Western blotting.

Protocol for sample preparation for cell lysis and efficient protein extraction from cultured tissues and cells for subsequent Western blotting.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service