Skip to Content
Merck
All Photos(2)

Key Documents

P9556

Sigma-Aldrich

N-(Phosphonomethyl)glycine

BioReagent, suitable for plant cell culture

Synonym(s):

Glyphosate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2P(O)CH2NHCH2CO2H
CAS Number:
Molecular Weight:
169.07
Beilstein:
2045054
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.71

product line

BioReagent

form

powder

technique(s)

cell culture | plant: suitable

mp

230 °C (dec.) (lit.)

application(s)

agriculture

shipped in

ambient

SMILES string

OC(=O)CNCP(O)(O)=O

InChI

1S/C3H8NO5P/c5-3(6)1-4-2-10(7,8)9/h4H,1-2H2,(H,5,6)(H2,7,8,9)

InChI key

XDDAORKBJWWYJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glyphosate (N-(phosphonomethyl)glycine) (sold commercially as Roundup) is the most widely used broad-spectrum systemic herbicide in the world. Consequently, the environmental effects of glyphosphate and its mechanism of action are an important area of active research.

Biochem/physiol Actions

Glyphosate (N-[phosphonomethyl] glycine) is the herbicide form of the isopropylamine salt of glyphosate.
Glyphosphate inhibits the shikimate pathway-dependent synthesis of the aromatic amino acids tyrosine, tryptophan and phenylalanine at the growing points of plants.

Preparation Note

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of glyphosate (N-(phosphonomethyl) glycine) on performance and selected parameters in broilers
Kubena L F, et al.
Poultry Science, 60(1), 132-136 (1981)
Apoorva Ravishankar et al.
Environmental pollution (Barking, Essex : 1987), 262, 114359-114359 (2020-05-24)
Glyphosate-based herbicides, the most extensively used herbicides in the world, are available in an enormous number of commercial formulations with varying additives and adjuvants. Here, we study the effects of one such formulation, Credit41, in two genetically diverse yeast strains.
Hugh J Beckie
Pest management science, 67(9), 1037-1048 (2011-05-07)
This review focuses on proactive and reactive management of glyphosate-resistant (GR) weeds. Glyphosate resistance in weeds has evolved under recurrent glyphosate usage, with little or no diversity in weed management practices. The main herbicide strategy for proactively or reactively managing
Stephen O Duke
Journal of agricultural and food chemistry, 59(11), 5835-5841 (2010-10-06)
High levels of aminomethylphosphonic acid (AMPA), the main glyphosate metabolite, have been found in glyphosate-treated, glyphosate-resistant (GR) soybean, apparently due to plant glyphosate oxidoreductase (GOX)-like activity. AMPA is mildly phytotoxic, and under some conditions the AMPA accumulating in GR soybean
Pedro J Christoffoleti et al.
Pest management science, 64(4), 422-427 (2008-02-20)
South America represents about 12% of the global land area, and Brazil roughly corresponds to 47% of that. The major sustainable agricultural system in South America is based on a no-tillage cropping system, which is a worldwide adopted agricultural conservation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service