Skip to Content
Merck
All Photos(5)

Key Documents

P7643

Sigma-Aldrich

PIPES dipotassium salt

≥99% (titration)

Synonym(s):

1,4-Piperazinediethanesulfonic acid dipotassium salt, Piperazine-1,4-bis(2-ethanesulfonic acid) dipotassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16K2N2O6S2
CAS Number:
Molecular Weight:
378.55
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥99% (titration)

form

crystalline powder

useful pH range

6.1-7.5

pKa (25 °C)

6.8

solubility

water: 0.25 g/mL, clear, colorless

application(s)

diagnostic assay manufacturing

SMILES string

[K+].[K+].[O-]S(=O)(=O)CCN1CCN(CC1)CCS([O-])(=O)=O

InChI

1S/C8H18N2O6S2.2K/c11-17(12,13)7-5-9-1-2-10(4-3-9)6-8-18(14,15)16;;/h1-8H2,(H,11,12,13)(H,14,15,16);;/q;2*+1/p-2

InChI key

BJQYFCAMUXGZFN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

PIPES (1,4-Piperazinediethanesulfonic acid) is a zwitterionic buffer. It is a stable buffer and is used majorly in fixation solutions. PIPES is inert towards enzymatic and non-enzymatic reactions. However, it generates artefacts in transmission electron microscope imaging.

Application

PIPES (1,4-Piperazinediethanesulfonic acid) dipotassium salt has been used as buffer for fluorescence polarization microscopy studies of kinesin-microtubule binding. It is suitable for use as a component of PHEM buffer ([K-PIPES], HEPES, EGTA, and MgSO4) used during rinsing and blocking steps in immunofluorescence imaging of HeLa cells. It is also suitable for use as buffer for purification of tubulin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of single molecule fluorescence polarization microscopy to study protein conformation and dynamics of kinesin-microtubule complexes
Benoit MPMH and Sosa H
Single Molecule Analysis, 15(1), 199-216 (2018)
Imaging the Drosophila retina: zwitterionic buffers PIPES and HEPES induce morphological artifacts in tissue fixation
Nie J, et al.
BMC Developmental Biology, 15(1), 10-10 (2015)
Dynamic phosphorylation of NudC by Aurora B in cytokinesis
Weiderhold KN
PLoS ONE, 11(4), e0153455-e0153455 (2016)
Helical alignment inversion of microtubules in accordance with a structural change in their lattice
Shikinaka K, et al.
Soft Matter, 11(19), 3869-3874 (2015)
Nina Marie Pedersen et al.
The Journal of cell biology, 219(8) (2020-06-02)
Cancer cells break tissue barriers by use of small actin-rich membrane protrusions called invadopodia. Complete invadopodia maturation depends on protrusion outgrowth and the targeted delivery of the matrix metalloproteinase MT1-MMP via endosomal transport by mechanisms that are not known. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service