Skip to Content
Merck
All Photos(1)

Key Documents

P5911

Sigma-Aldrich

1,2-Dipalmitoyl-rac-glycero-3-phosphocholine

~99%

Synonym(s):

DL-α-Phosphatidylcholine, dipalmitoyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H80NO8P
CAS Number:
Molecular Weight:
734.04
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Assay

~99%

Quality Level

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OCC(COP(O)(=O)OCC[N](C)(C)C)OC(=O)CCCCCCCCCCCCCCC

InChI

1S/C40H81NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-39(42)46-36-38(37-48-50(44,45)47-35-34-41(3,4)5)49-40(43)33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h38H,6-37H2,1-5H3,(H,44,45)

InChI key

RMBFBWDOAGQYBH-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katherine C Thompson et al.
Langmuir : the ACS journal of surfaces and colloids, 29(14), 4594-4602 (2013-03-14)
The presence of unsaturated lipids in lung surfactant is important for proper respiratory function. In this work, we have used neutron reflection and surface pressure measurements to study the reaction of the ubiquitous pollutant gas-phase ozone, O3, with pure and
Michelle L Gee et al.
Scientific reports, 3, 1557-1557 (2013-03-28)
Antimicrobial peptides hold promise as broad-spectrum alternatives to conventional antibiotics. The mechanism of action of this class of peptide is a topical area of research focused predominantly on their interaction with artificial membranes. Here we compare the interaction mechanism of
Julio C P Damalio et al.
Biochimica et biophysica acta, 1828(6), 1441-1448 (2013-02-19)
The molecular mechanisms responsible for protein structural changes in the central nervous system leading to Alzheimer's disease are unknown, but there is evidence that a family of proteins known as septins may be involved. Septins are a conserved group of
Jorge Bernardino de la Serna et al.
Faraday discussions, 161, 535-548 (2013-06-29)
Pulmonary surfactant is a lipid-protein complex essential to stabilize alveoli, by forming surface active films able to reach and sustain very low surface tensions (< 2 mN m(-1)) during the film compression that occurs at end-expiration. The particular lipid composition
Zeynep Busra Bolat et al.
Frontiers in bioengineering and biotechnology, 8, 50-50 (2020-03-03)
Combination chemotherapy, administrating two chemotherapeutic agents concurrently, comes into prominence, as the heterogeneity or the level of the disease necessitates a collaborative action. Curcumin, isolated from turmeric, and piperine, isolated from black long pepper, are two dietary polyphenols studied for

Articles

Read our article about how the Nanodisc system allows for structural studies of membrane proteins.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service