P4691
Protein G-Agarose, Fast Flow from Streptococcus sp.
saline suspension
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
41106500
Recommended Products
form
saline suspension
extent of labeling
≥2 mg per mL
matrix
Highly cross-linked 4% beaded agarose, fast flow
matrix activation
cyanogen bromide
matrix attachment
amino
matrix spacer
1 atom
capacity
≥20 mg/mL binding capacity (human IgG (I 4506))(in 20 mM sodium phosphate, pH 7.0)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Protein G-agarose is used in affinity chromatography, protein chromatography, antibody purification and characterization, immunoaffinity matrices, protein A, G and L resins, and purification and detection. Protein G-agarose has been used to study breast cancer and falsely elevated thyroid-stimulating hormone (TSH).
Physical form
Suspension in 0.5 M NaCl containing 20% ethanol as a preservative
Preparation Note
Prepared with a genetically engineered Protein G which retains its high affinity for IgG and lacks albumin and Fab binding sites and membrane binding regions.
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Cheng Song et al.
Journal of cellular and molecular medicine, 20(11), 2064-2077 (2016-10-27)
Interleukin (IL)-1β plays an important role in the pathogenesis of idiopathic pulmonary fibrosis. The production of IL-1β is dependent upon caspase-1-containing multiprotein complexes called inflammasomes and IL-1R1/MyD88/NF-κB pathway. In this study, we explored whether a potential anti-fibrotic agent fluorofenidone (FD)
J T Lin et al.
Molecular and cellular biology, 19(5), 3237-3245 (1999-04-17)
Upon heat shock, transcription of many stress-inducible genes is rapidly and dramatically stimulated by heat shock factor (HSF). A central region of the yeast HSF (designated HSFrr for "repression region") was previously identified and proposed to be involved in repressing
L Zahradova et al.
Neoplasma, 59(4), 440-449 (2012-04-12)
In a phase II clinical study, pretreated multiple myeloma patients with relapsing or stable disease received autologous anticancer vaccine containing dendritic cells loaded with Id-protein. Patients received a total of 6 vaccine doses intradermally in monthly intervals. No clinical responses were observed.
Fan Jiang et al.
The Journal of pharmacology and experimental therapeutics, 324(1), 261-269 (2007-10-06)
Previously we have demonstrated that 3',4'-dihydroxyflavonol (DiOHF), a novel synthetic flavonol, protects against ischemia reperfusion injury in both heart and brain. In this study, we characterized the pharmacological effects of DiOHF on phagocytic and vascular NADPH oxidase. Superoxide release (lucigenin-enhanced
Marek Babusiak et al.
Proteomics, 7(1), 121-129 (2007-01-06)
Intestinal epithelial cell protrusions referred as microvilli or brush border membranes (BBMs) are specialized in the digestion, uptake, and transport of nutrients, trace elements and vitamins from intestinal lumen into the circulation. Disorders of intestinal absorption are common in human
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service