Skip to Content
Merck
All Photos(1)

Key Documents

P4649

Sigma-Aldrich

Phosphorylase b from rabbit muscle

For use as a marker in SDS-PAGE

Synonym(s):

α-Glucan Phosphorylase, 1,4-α-D-Glucan:orthophosphate α-D-glucosyltransferase, Glycogen Phosphorylase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

powder

mol wt

observed mol wt ~97 kDa

packaging

vial of 0.5 mg

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Phosphorylase b is a dimer, usually exists in inactive form in the skeletal muscles. Equilibrium exists between an active relaxed (R) state and a less active tense (T). Phosphorylase b favors the T state. The enzyme possesses three domains, the N-terminal domain, glycogen-binding domain and the C-terminal domain.

Application

Phosphorylase b from rabbit muscle has been used as a molecular weight marker in 12% polyacrylamide gel for keratinase, and stress-70 protein.
Phosphorylase b from rabbit muscle is to be used as a marker in SDS-PAGE. Phosphorylase b is used during chemical cross-linking studies as a SDS-PAGE molecular weight standard.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystallographic studies on the activity of glycogen phosphorylase b
Weber IT, et al.
Nature, 274(5670), 433-433 (1978)
Signal Transduction and Protein Phosphorylation (2012)
Stress-70 proteins in marine mussel Mytilus galloprovincialis as biomarkers of environmental pollution: a field study
Hamer B, et al.
Environment International, 30(7), 873-882 (2004)
B Chen et al.
The Journal of biological chemistry, 275(45), 34946-34953 (2000-08-17)
The envelope glycoprotein, gp160, of simian immunodeficiency virus (SIV) shares approximately 25% sequence identity with gp160 from the human immunodeficiency virus, type I, indicating a close structural similarity. As a result of binding to cell surface CD4 and co-receptor (e.g.
Phosphorylase Is Regulated by Allosteric Interactions and Reversible Phosphorylation
Berg JM, et al.
Biochemistry (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service