Skip to Content
Merck
All Photos(1)

Documents

P3111

Sigma-Aldrich

Protease from Bacillus sp.

liquid, ≥16 U/g

Synonym(s):

Savinase® 16.0L

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Bacillus sp.

form

liquid

specific activity

≥16 U/g

mol wt

20-30 kDa

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Protease is an enzyme used to break down proteins by hydrolyzing peptide bonds. Product P3111 is secreted from Bacillus sp.. It is a serine-type protease characterized by excellent performance at elevated temperature and pH. Product P3111 is extremely stable under harsh storage conditions, such as in the presence of sodium hypochlorite. Protease is used to degrade proteins, to study protease inhibitors and to study thermal inactivation kinetics.

Biochem/physiol Actions

Protease catabolizes proteins by hydrolysis of peptide bonds. Proteases are inactivated by serine active-site inhibitors, such as phenylmethylsulfonyl fluoride (PMSF) and diisopropylfluorophosphate. Proteases, secreted from Bacillus sp., typically have molecular weights ranging from 20,000 to 30,000. They are typically stabilized by Ca2+ and have high isoelectric points. It is highly soluble in detergents at a wide range of pH and temperatures.

Legal Information

A product of Novozyme Corp.
Savinase is a registered trademark of Novozymes Corp.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessio Gomiero et al.
Environmental science and pollution research international, 26(24), 24407-24416 (2019-06-24)
In recent years, the occurrence of microplastics in the aquatic environment has gathered increasing scientific interest. Several studies have shown that the ingestion of microplastics may negatively influence the physiology of marine organisms having different feeding strategies, particularly in those
Cheng Wang et al.
mSystems, 5(1) (2020-02-13)
Substantial annual economic loss in livestock production is caused by antinutritional factors in soybean meal and corn mixed substrates, which can be degraded by microbial fermentation. Although considerable efforts have been made to explain the effects of fermentation on soybean
Yoshifumi Shimada et al.
Human pathology, 66, 1-9 (2017-02-27)
HER2-targeted therapy is considered effective for KRAS codon 12/13 wild-type, HER2-positive metastatic colorectal cancer (CRC). In general, HER2 status is determined by the use of immunohistochemistry (IHC) and fluorescence in situ hybridization (FISH). Comprehensive genomic sequencing (CGS) enables the detection
Weifa Su et al.
Frontiers in microbiology, 12, 688839-688839 (2021-09-07)
Corn germ meal (CGM) and corn gluten feed (CGF) are the two main corn byproducts (CBs) obtained from corn starch extraction. Due to their high fiber content, low protein content, and severe imbalance of amino acid, CBs are unable to
Juliana A S Leite et al.
Frontiers in nutrition, 8, 626475-626475 (2021-03-30)
Proteases present in milk are heat-sensitive, and their activities increase or decrease depending on the intensity of the thermal treatment applied. The thermal effects on the protease activity are well-known for bovine milk but poorly understood for ovine and caprine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service