Skip to Content
Merck
All Photos(1)

Key Documents

P2318

Sigma-Aldrich

Anti-Pseudomonas Exotoxin A antibody produced in rabbit

whole antiserum

Synonym(s):

Anti-ETA

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

(Pseudomonas aeruginosa)

technique(s)

dot blot: 1:20,000
indirect ELISA: 1:250,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Pseudomonas aeruginosa PAO1 ... toxA(877850)

General description

Reacts against Pseudomonas exotoxin A, but shows no reaction against Staphylococcal enterotoxin A, Cholera toxin, and Staphylococcal enterotoxin B. The product has not been tested for neutralization potency against active Pseudomonas exotoxin A.

Immunogen

exotoxin A from Pseudomonas aeruginosa.

Application

Anti-Pseudomonas Exotoxin A antibody has been used in in vitro serum stability of fusion toxins and binding of targeted toxins.
Anti-Pseudomonas exotoxin A antibody produced in rabbit has been used:
  • in the in vitro analysis of the stability of fusion toxins in serum
  • to construct an epidermal growth factor receptor (EGFR)-targeted toxin to study its toxicity on Swiss mouse embryo NIH-3T3 cells transfected with human EGFR
  • in the determination of the in vivo half-life of immunotoxins in serum

Biochem/physiol Actions

Pseudomonas exotoxin A is secreted by Pseudomonas aeruginosa and has potent cytotoxic effects. Exotoxin A enters the cells by receptor-mediated endocytosis, translocates to the cytoplasm and inhibits protein synthesis by ADP-ribosylating factor 2. Chimeric exotoxin A has applications in gene treatment of various diseases and probable treatment for cancer.

Physical form

This product is supplied as a liquid containing 15 mM sodium azide as preservative.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Weng et al.
Molecular oncology, 6(3), 323-332 (2012-02-09)
Tumor-targeting protein toxins are composed of a toxic enzyme coupled to a specific cell binding domain that targets cancer-associated antigens. The anti-tumor treatment by targeted toxins is accompanied by dose-limiting side effects. The future prospects of targeted toxins for therapeutic
Moritz Bewarder et al.
Cancer immunology, immunotherapy : CII, 69(8), 1535-1548 (2020-04-18)
With an infection rate of 60-90%, the human cytomegalovirus (HCMV) is very common among adults but normally causes no symptoms. When T cell-mediated immunity is compromised, HCMV reactivation can lead to increased morbidity and mortality. HCMV antigens are processed and
Fusion of an albumin-binding domain extends the half-life of immunotoxins
Guo R, et al.
International Journal of Pharmaceutics, 511(1), 538-549 (2016)
Alexander N Wein et al.
Journal of medicinal chemistry, 55(18), 7998-8006 (2012-09-08)
Protective antigen (PA), lethal factor, and edema factor, the protein toxins of Bacillus anthracis , are among its most important virulence factors and play a key role in infection. We performed a virtual ligand screen of a library of 10000
The toxin component of targeted anti-tumor toxins determines their efficacy increase by saponins
Weng A, et al.
Molecular Oncology, 6(3), 323-332 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service