Skip to Content
Merck
All Photos(4)

Documents

P2172

Sigma-Aldrich

C-Phycocyanin from Spirulina sp.

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.56

biological source

Spirulina

Quality Level

form

lyophilized powder

composition

Protein, ~40% Lowry

concentration

30-50% protein (Lowry)

storage temp.

2-8°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

C-Phycocyanin is a protein pigment, that is soluble in water. It is an important constituent of Spirulina platensis.

Application

C-Phycocyanin from Spirulina sp. has been used:
  • as a standard to characterize the fluorescence interactions and oxidation behaviors for model compounds
  • as a standard to confirm the antibacterial activity
  • to characterize the recombinantly expressed α-subunit of C-phycocyanin from Synechocystis sp. PCC6803 (HisCPC)
  • to identify the phycocyanin from water extracted Spirulina maxima sample

Biochem/physiol Actions

C-phycocyanin, a biliprotein with antioxidant properties, was shown to induce apoptosis in HeLa cells by stimulating expression of pro-apoptotic genes as well as by inducing increased release of cytochrome c from the mitochondria.
C-phycocyanin can act as an active inhibitor of A53Tα-synuclein. It possess anti-inflammatory properties.

Packaging

Package size based on protein content

Physical form

Partially purified, contains sucrose, dithioerythritol and sodium azide as preservatives

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C-Phycocyanin from Spirulina Inhibits ?-Synuclein and Amyloid ?-Fibril Formation but Not Amorphous Aggregation
Liu Y, et al.
Journal of Natural Products, 82(1), 66-73 (2019)
C-phycocyanin: a potent peroxyl radical scavenger in vivo and in vitro
Bhat VB and Madyastha KM
Biochemical and biophysical research communications, 275(1), 20-25 (2000)
Evaluating fluorescence spectroscopy as a tool to characterize cyanobacteria intracellular organic matter upon simulated release and oxidation in natural water
Korak JA, et al.
Water Research, 68(1), 432-443 (2015)
Bing Li et al.
Biotechnology and applied biochemistry, 43(Pt 3), 155-164 (2005-12-01)
C-phycocyanin (C-PC), a water-soluble protein pigment, isolated from Spirulina platensis, is of great importance because of its various medical and pharmacological properties. In the present study, we first investigated the effect of highly purified C-PC on growth and proliferation of
Protective effect of water extracted Spirulina maxima on glutamate-induced neuronal cell death in mouse hippocampal HT22 cell
Lee HY, et al.
Pharmacognosy magazine, 14(54), 242-242 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service