Skip to Content
Merck
All Photos(1)

Key Documents

P1132

Sigma-Aldrich

Pterine

~95%

Synonym(s):

2-Amino-4-hydroxypteridine, 2-Amino-4-oxodihydropteridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5N5O
CAS Number:
Molecular Weight:
163.14
Beilstein:
150294
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Assay

~95%

form

powder

storage temp.

2-8°C

SMILES string

Nc1nc(O)c2nccnc2n1

InChI

1S/C6H5N5O/c7-6-10-4-3(5(12)11-6)8-1-2-9-4/h1-2H,(H3,7,9,10,11,12)

InChI key

HNXQXTQTPAJEJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Pterine has been used:
  • in photochemicals assay for inducing DNA damage
  • as a substrate in xanthine oxidase assay
  • as a standard in thin layer chromatography (TLC) for quantifying pterine from microbes

Biochem/physiol Actions

Pterine consists of pyrazine and pyrimidine with substitutions in keto and amino group. Pterine is present in all organisms including cyanobacteria. It acts as a cofactor for redox enzymes and serves as a precursor for folic acid synthesis. Pterine from the butterfly wings absorb light and elicit incoherent scattering. Pterine induces alterations in DNA by hydroxylation of deoxyguanosine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pterin function in bacteria
Feirer N and Fuqua C
Pteridines, 18(1), 23-36 (2017)
Andrea Sariego-Jamardo et al.
Pediatric neurology, 53(5), 422-426 (2015-10-18)
The mechanisms of the ketogenic diet remain unclear, but several predictors of response have been proposed. We aimed is to study the relationship between the etiology of epilepsy, cerebrospinal fluid neurotransmitters, pterins, and amino acids, and response to a ketogenic
Febuxostat, an inhibitor of xanthine oxidase, suppresses lipopolysaccharide-induced MCP-1 production via MAPK phosphatase-1-mediated inactivation of JNK
Nomura J, et al.
PLoS ONE, 8(9), e75527-e75527 (2013)
Evaluation of Pterin, a Promising Drug Candidate from Cyanide Degrading Bacteria
Mahendran R, et al.
Current Microbiology, 75(6), 684-693 (2018)
Photoinduced cleavage of plasmid DNA in the presence of pterin
Lorente C, et al.
Pteridines, 11(3), 100-105 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service