Skip to Content
Merck
All Photos(1)

Key Documents

P0194

Sigma-Aldrich

Protein Kinase Cζ isozyme human

≥75% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, buffered aqueous solution

Synonym(s):

Ca2+-activated phospholipid-dependent serine-threonine kinase ζ isozyme human, PKCζ human

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in baculovirus infected insect cells

Quality Level

Assay

≥75% (SDS-PAGE)

form

buffered aqueous solution

enzyme activity

>800 units/mg protein

mol wt

76-80 kDa by SDS-PAGE

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PRKCZ(5590)

Biochem/physiol Actions

Protein Kinase C (PKC) is a serine/threonine kinase that is activated intracellularly by signal transduction pathways that produce DAG from phosphatidylinositol diphosphate (PIP2) and phosphatidylcholine (PC) through the action of various activated phospholipases. Phorbol esters also stimulate PKC. At least 11 PKC isozymes have been identified that differ in primary structure, tissue distribution, subcellular localization, response to extracellular signals, and substrate specificity. The isozymes can be grouped into three subfamilies. Members of the first family require Ca2+ and phospholipid and include PKCα, βI, βII, and γ. Members of the second family are phospholipid-dependent but Ca2+-independent, and include PKCδ, ε, η, and θ. Members of the third family are not activated by either DAG or phorbol esters and include PKCξ, μ, and ι.
Phosphorylation appears to be an important mechanism of regulation of all PKCs. PKC plays a role in the regulation of cell transformation, growth, differentiation, ruffling, vesicle trafficking, apoptosis and gene expression.

Suitability

PKC ε can transfer 1100 nmole of phosphate to PKC ε substrate peptide per minute per mg of total protein at 30 °C.

Unit Definition

One unit will transfer 1 nanomole of phosphate to PKC epsilon substrate peptide per minute at pH 7.5 at 30 deg C.

Physical form

Solution in 20 mM HEPES, pH 7.5; 2 mM EDTA, 2 mM EGTA, 5 mM DTT, 250 mM NaCl, 0.05% Triton X-100, and 50% glycerol.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Girdhar K Pandey et al.
Cell research, 17(5), 411-421 (2007-05-09)
Potassium is one of the major macro-nutrients essential for a number of cellular processes in plants. Low potassium level in the soil represents a limiting factor for crop production. Recent studies have identified potassium transporters that are involved in potassium
Yong Xiang et al.
Plant physiology, 144(3), 1416-1428 (2007-05-31)
Plants respond to adverse environments by initiating a series of signaling processes that often involves diverse protein kinases, including calcineurin B-like protein-interacting protein kinases (CIPKs). In this study, putative CIPK genes (OsCIPK01-OsCIPK30) in the rice (Oryza sativa) genome were surveyed
Cecilia D'Angelo et al.
The Plant journal : for cell and molecular biology, 48(6), 857-872 (2006-11-10)
Intracellular release of calcium ions belongs to the earliest events in cellular stress perception. The molecular mechanisms integrating signals from different environmental cues and translating them into an optimized response are largely unknown. We report here the functional characterization of
Jun Hou et al.
Journal of thoracic oncology : official publication of the International Association for the Study of Lung Cancer, 7(1), 105-114 (2011-12-03)
A challenge of cancer therapy is to optimize therapeutical options to individual patients. Cancers with similar histology may show dramatically different responses to therapy, indicating that a refined approach needs to be developed to classify tumors by intrinsic characteristics that
Hai Huang et al.
Development (Cambridge, England), 138(12), 2477-2485 (2011-05-13)
Post-translational modification by the small ubiquitin-related modifier (SUMO) is important for a variety of cellular and developmental processes. However, the precise mechanism(s) that connects sumoylation to specific developmental signaling pathways remains relatively less clear. Here, we show that Smt3 knockdown

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service