Skip to Content
Merck
All Photos(1)

Key Documents

O7635

Sigma-Aldrich

Anti-Osteopontin antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-OPN

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

capture ELISA: 0.2-0.8 μg/mL
immunohistochemistry: 5-15 μg/mL using mouse thymus tissue sections
neutralization: 1-3 μg/mL
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Spp1(20750)

General description

The OPN (osteopontin) gene is mapped to mouse chromosome 5 E5 and codes for secreted phosphoprotein 1 and is mostly expressed in bone tissue.

Immunogen

purified recombinant mouse osteopontin expressed in NSO cells.

Biochem/physiol Actions

Osteopontin (OPN) serves as a ligand to a number of integrin receptors which are associated with cell motility, chemotaxis, invasion and cell survival. Upregulation of OPN gene is observed in tumors such as colorectal carcinomas, breast cancer and contributes to its pathogenesis. OPN is associated with obesity by mediating adipocyte inflammation and insulin resistance upon macrophage activation. OPN functions as an effector molecule in extracellular signalling and promotes fibrosis and myocyte growth.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Analysis Note

Neutralizes the biological activity of recombinant mouse osteopontin using 293 cells.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suppression of osteopontin inhibits chemically induced hepatic carcinogenesis by induction of apoptosis in mice.
Lee S H, et al.
Oncotarget, 7(52), 87219-87219 (2016)
Osteopontin Deficiency Suppresses Intestinal Tumor Development in Apc-Deficient Min Mice.
Ishigamori R, et al.
International Journal of Molecular Sciences, 18(5), 1058-1058 (2017)
Tracking the embryonic stem cell transition from ground state pluripotency.
Kalkan T, et al.
Development, dev-142711 (2017)
Transcriptome analysis of fetal metatarsal long bones by microarray, as a model for endochondral bone formation.
Sugars R V, et al.
Biochimica et Biophysica Acta (BBA)-Gene Structure and Expression, 1763(10), 1031-1039 (2006)
Mio Yamane et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(8), 10778-10800 (2020-07-04)
Chronic graft-vs-host disease (cGVHD) is a multifactorial inflammatory disease that affects patients undergoing hematopoietic stem cell transplantation. Multiple organs, including the lacrimal glands (LGs), are negatively affected by cGVHD and lose function due to the resultant fibrosis. An abnormal immune

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service